Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • GABRA1 Antibodies

          Antibodies Inc

          GABA-A-R/Alpha1 Monoclonal Antibody (N95/35)

          View all (31) GABRA1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GABA-A-R/Alpha1 Monoclonal Antibody (N95/35)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          GABA-A-R/Alpha1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GABA-A-R/Alpha1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GABA-A-R/Alpha1 Antibody (75-136) in ICC/IF

          The distribution of both alpha-1 (Product # 75-136, 1:1000; green) and alpha-6 (red) subunits was investigated on the cerebellar granule cells. Both subunits were each co-stained with the inhibitory GABAergic presynaptic neuronal marker protein vGAT (blue). alpha-6 immunoactivity was present along the neurite and alpha-1 was predominately present in synapses opposing the pre-synaptic vGAT (E), where alpha-6... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GABA-A-R/Alpha1 Antibody in Immunocytochemistry (ICC/IF)
          GABA-A-R/Alpha1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GABA-A-R/Alpha1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GABA-A-R/Alpha1 Antibody in Western Blot (WB)
          GABA-A-R/Alpha1 Antibody in Western Blot (WB)
          GABA-A-R/Alpha1 Monoclonal Antibody (N95/35)

          Product Details

          75-136

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:250
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Xenopus

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N95/35

          Immunogen

          Fusion protein amino acids 355-394 of mouse GABA-AR-Alpha1 (accession number P62812) produced recombinantly in E. Coli.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4

          Contains

          0.065% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          Gamma -aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date aix alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha- and beta-subunits results in the expression of functional GABAA-R sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta-subunits of the receptor has been shown to modulate GABAA-R function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GABA(A) receptor subunit alpha-1; GABA(A) receptor, alpha 1; GABAA alpha 1; GABAAR alpha1; GABAAR subunit alpha-1; GABRA 1; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, alpha 1; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 1; gamma-aminobutyric acid A receptor, alpha 1; Gamma-aminobutyric acid receptor subunit alpha-1; gamma-aminobutyric acid type A receptor alpha1 subunit; unnamed protein product

          View more View less

          Gene Aliases: DEE19; ECA4; EIEE19; EJM; EJM5; Gabra-1; GABRA1

          View more View less

          UniProt ID: (Human) P14867, (Rat) P62813, (Mouse) P62812

          View more View less

          Entrez Gene ID: (Human) 2554, (Rat) 29705, (Mouse) 14394

          View more View less

          Function(s)
          transmembrane signaling receptor activity GABA-A receptor activity ion channel activity extracellular ligand-gated ion channel activity chloride channel activity benzodiazepine receptor activity GABA receptor activity GABA-gated chloride ion channel activity signaling receptor activity transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential inhibitory extracellular ligand-gated ion channel activity protein binding drug binding ligand-gated ion channel
          Process(es)
          ion transport chloride transport gamma-aminobutyric acid signaling pathway ion transmembrane transport synaptic transmission, GABAergic regulation of postsynaptic membrane potential chloride transmembrane transport inhibitory synapse assembly cellular response to histamine transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline