Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • GABRB3 Antibodies

          Bioss

          GABR B3 Polyclonal Antibody

          View all (22) GABRB3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GABR B3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          GABR B3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GABR B3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GABR B3 Antibody (BS-8544R) in WB

          Lane 1: Mouse Cerebrum, Lane 2: Mouse Hippocampus; Probed with GABRB3 Polyclonal Antibody, unconjugated (bs-8544R) at 1:1000 overnight at 4°C followed by a conjugated secondary antibody at 1:20000 for 90 minutes at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GABR B3 Antibody in Western Blot (WB)
          GABR B3 Antibody in Western Blot (WB)
          GABR B3 Polyclonal Antibody

          Product Details

          BS-8544R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:400
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          Flow Cytometry (Flow)

          1:20-1:100
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human GABR B3/GABA A Receptor beta 3, amino acids 31-130.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Gamma-amino butyric acid (GAMA) is a strong inhibitor of neuron transmission and is found throughout the central nervous system. GABA is formed by the decarboxylation of glutamate, resulting in the formation of one of two distinct types; GABA-A or GABA-B.The binding of GABA to GABA-A receptors results in the increase transportation of chlorine ions into the neuron. This influx results in signal transmission repression. Studies have shown that if cell to cell signaling goes unchecked due to a lack of GABA, a system overload can resulting in blackouts, rapid body contractions, seizures and epilepsy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cleft palate 1; GABA(A) receptor subunit beta-3; GABA-alpha receptor beta-2 subunit; GABAA receptor beta-3 subunit; GABAAR subunit beta-3; GABR B3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor beta 3; Gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; gamma-aminobutyric acid type A receptor beta3 subunit; gamma-aminobutyric acid type A receptor subunit beta 3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3; unnamed protein product

          View more View less

          Gene Aliases: A230092K12Rik; AW049585; beta3; Cp1; DEE43; ECA5; EIEE43; Gabrb-3; GABRB3

          View more View less

          UniProt ID: (Human) P28472, (Mouse) P63080, (Rat) P63079

          View more View less

          Entrez Gene ID: (Human) 2562, (Mouse) 14402, (Rat) 24922

          View more View less

          Function(s)
          transmembrane signaling receptor activity GABA-A receptor activity ion channel activity extracellular ligand-gated ion channel activity chloride channel activity GABA-gated chloride ion channel activity identical protein binding transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential AP-2 adaptor complex binding protein binding ligand-gated ion channel
          Process(es)
          ion transport chloride transport signal transduction gamma-aminobutyric acid signaling pathway chemical synaptic transmission nervous system development brain development learning memory response to xenobiotic stimulus reproductive behavior cerebellum development ion transmembrane transport social behavior exploration behavior circadian sleep/wake cycle, REM sleep neuron development synaptic transmission, GABAergic palate development hard palate development inhibitory postsynaptic potential inner ear receptor cell development innervation motor behavior cellular response to zinc ion cellular response to histamine response to anesthetic cochlea development chloride transmembrane transport inhibitory synapse assembly transport sensory perception of sound regulation of neuron apoptotic process negative regulation of neuron apoptotic process negative regulation of neuron death
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline