Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GAK Antibodies

          Abnova

          GAK Polyclonal Antibody, MaxPab™

          View all (13) GAK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GAK Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          GAK Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GAK Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GAK Antibody (H00002580-D01P) in WB

          Western Blot analysis of GAK expression in transfected 293T cell line (H00002580-T01) by GAK MaxPab polyclonal antibody.

          Lane 1: GAK transfected lysate(44.00 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GAK Antibody in Western Blot (WB)
          GAK Antibody in Western Blot (WB)
          GAK Polyclonal Antibody, MaxPab™

          Product Details

          H00002580-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GAK (AAH08668.1, 1 a.a. approximately 416 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MSLLQSALDF LAGPGSLGGA SGRDQSDFVG QTVELGELRL LLASPAPPLS VQSTPRGGPP AAADPFGPLL PSSGNNSQPC SNPDLFGEFL NSDSVTVPPS FPSAHSSPPP SCSADFLHLG DLPGEPSKMT ASSSNPDLLG GWAAWTETAA SAVAPTPATE GPLFSPGGQP APCGSQASWT KSQNPDPFAD LGDLSSGLQG SPAGFPPGGF IPKTATTPKG SSSWQTSRPP AQGASWPPQA KPPPKACTQP RPNYASNFSV IGAREERGVR APSFAQKPKV SENDFEDLLS NQGFSSRSDK KGPKTIAEMR KQDLAKDTDP LKLKLLDWIE GKERNIRALL STLHTVLWDG ESRWTPVGMA DLVAPEQVKK HYRRAVLAVH PDKAAGQPYE QHAKMIFMEL NDAWSEFENQ GSRPLF

          Target Information

          GAK serine/threonine kinase or Cyclin-G-associated kinase was originally identified as a protein that is associated with cyclin G. However, neither CDK5 nor GAK kinase activity is activated by cyclin G nor does GAK affect CDK5 kinase activity. GAK seems to act as an auxilin homolog that is involved in the uncoating of clathrin-coated vesicles by Hsc70 in non-neuronal cells. Expression oscillates slightly during the cell cycle, peaking at G1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: auxilin-2; Cyclin-G-associated kinase; DnaJ homolog subfamily C member 26; unnamed protein product

          View more View less

          Gene Aliases: D130045N16Rik; DNAJ26; DNAJC26; GAK

          View more View less

          UniProt ID: (Human) O14976, (Mouse) Q99KY4

          View more View less

          Entrez Gene ID: (Human) 2580, (Mouse) 231580

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity clathrin binding cyclin binding protein binding involved in protein folding protein serine kinase activity transferase activity, transferring phosphorus-containing groups membrane traffic protein
          Process(es)
          protein folding receptor-mediated endocytosis endoplasmic reticulum organization Golgi organization negative regulation of neuron projection development synaptic vesicle uncoating protein localization to Golgi apparatus intracellular transport clathrin coat assembly clathrin coat disassembly clathrin-mediated endocytosis protein localization to plasma membrane Golgi to lysosome transport regulation of clathrin coat assembly protein phosphorylation cell cycle epidermal cell differentiation phosphorylation intrahepatic bile duct development cell development forebrain morphogenesis establishment of skin barrier positive regulation of neural precursor cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline