Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MLCCMRRTK QVEKNDDDQK IEQDGIKPED KAHKAATKIQ ASFRGHITRK KLKGEKKDDV QAAEAEANKK DEAPVADGVE KKGEGTTTAE AAPATGSKPD EPGKAGETPS EEKKGEGDAA TEQAAPQAPA SSEEKAGSAE TESATKASTD NSPSSKAEDA PAKEEPKQAD VPAAVTAAAA TTPAAEDAAA KATAQPPTET GESSQAEENI EAVDETKPKE SARQDEGKEE EPEADQEHA (1-238 aa encoded by BC007936 )
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile "growth cones" that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axonal membrane protein GAP-43; brain abundant, membrane attached signal protein 2; calmodulin-binding protein; calmodulin-binding protein P-57; GAP-43 protein; growth accentuating protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; neuron growth-associated protein 43; neuron protein F1; neuronal growth factor 43; protein F1; unnamed protein product
Gene Aliases: B-50; Basp2; GAP-43; PP46
UniProt ID: (Human) P17677, (Mouse) P06837, (Rat) P07936
Entrez Gene ID: (Human) 2596, (Mouse) 14432, (Rat) 29423
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support