Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • GAS6 Antibodies

          Bioss

          GAS 6 Polyclonal Antibody

          View all (10) GAS6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GAS 6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          GAS 6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GAS 6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GAS 6 Antibody (BS-7549R) in IHC (P)

          Formalin-fixed and paraffin embedded human lung carcinoma labeled with Rabbit Anti-GAS 6 Polyclonal Antibody, Unconjugated (bs-7549R) at 1:200 followed by conjugation to the secondary antibody and DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GAS 6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GAS 6 Antibody in Western Blot (WB)
          GAS 6 Polyclonal Antibody

          Product Details

          BS-7549R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human GAS 6, amino acids 551-650.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AXL receptor tyrosine kinase ligand; AXL stimulatory factor; GAS 6; GAS-6; GPF; Growth arrest-specific protein 6; growth-potentiating factor; unnamed protein product

          View more View less

          Gene Aliases: AXLLG; AXSF; Gas-6; GAS6

          View more View less

          UniProt ID: (Human) Q14393, (Mouse) Q61592

          View more View less

          Entrez Gene ID: (Pig) 100519331, (Human) 2621, (Rat) 58935, (Mouse) 14456

          View more View less

          Function(s)
          phosphatidylserine binding receptor binding calcium ion binding protein binding protein binding, bridging receptor tyrosine kinase binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process metal ion binding receptor agonist activity voltage-gated calcium channel activity growth factor activity protein tyrosine kinase activator activity binding, bridging extracellular matrix protein
          Process(es)
          neuron migration positive regulation of protein phosphorylation positive regulation of cytokine-mediated signaling pathway positive regulation of glomerular filtration protein phosphorylation phagocytosis apoptotic process signal transduction cell surface receptor signaling pathway enzyme linked receptor protein signaling pathway blood coagulation cellular response to starvation positive regulation of gene expression negative regulation of tumor necrosis factor-mediated signaling pathway cell migration fusion of virus membrane with host plasma membrane viral genome replication cell differentiation organ regeneration cell-substrate adhesion cellular response to nutrient levels positive regulation of TOR signaling activation of protein kinase B activity negative regulation of interferon-gamma production negative regulation of interleukin-1 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production positive regulation of natural killer cell differentiation myeloid cell apoptotic process negative regulation of myeloid cell apoptotic process cellular response to interferon-alpha B cell chemotaxis negative regulation of apoptotic process apoptotic cell clearance fibroblast apoptotic process positive regulation of protein kinase activity negative regulation of transcription, DNA-templated viral entry into host cell receptor-mediated virion attachment to host cell positive regulation of protein export from nucleus positive regulation of fibroblast proliferation positive regulation of phagocytosis positive regulation of protein kinase B signaling negative regulation of biomineral tissue development positive regulation of ERK1 and ERK2 cascade calcium ion transmembrane transport cellular response to vitamin K cellular response to glucose stimulus cellular response to growth factor stimulus cellular response to xenobiotic stimulus protein localization to plasma membrane extracellular matrix assembly dendritic cell differentiation hematopoietic stem cell migration to bone marrow negative regulation of oligodendrocyte apoptotic process negative regulation of fibroblast apoptotic process negative regulation of endothelial cell apoptotic process positive regulation of dendritic cell chemotaxis negative regulation of renal albumin absorption negative regulation of dendritic cell apoptotic process macrophage cytokine production peptidyl-serine phosphorylation positive regulation of peptidyl-serine phosphorylation negative regulation of protein import into nucleus, translocation cellular response to drug regulation of growth tissue regeneration negative regulation of cysteine-type endopeptidase activity involved in apoptotic process negative regulation of sequence-specific DNA binding transcription factor activity protein kinase B signaling tissue remodeling negative regulation of interleukin-1 secretion positive regulation of protein tyrosine kinase activity protein targeting to plasma membrane negative regulation of interleukin-6 secretion positive regulation of receptor activity regulation of ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline