Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • GAS2L2 Antibodies

          Abnova

          GAS2L2 Polyclonal Antibody, MaxPab™

          View all (2) GAS2L2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GAS2L2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          GAS2L2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GAS2L2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GAS2L2 Antibody (H00246176-B01P) in WB

          Western Blot analysis of GAS2L2 expression in transfected 293T cell line by GAS2L2 MaxPab polyclonal antibody. Lane 1: GAS2L2 transfected lysate(96.8 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GAS2L2 Antibody in Western Blot (WB)
          GAS2L2 Polyclonal Antibody, MaxPab™

          Product Details

          H00246176-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GAS2L2 (NP_644814.1, 1 a.a. approximately 880 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSQPAGGRRK PRTLGPPVCS IRPFKSSEQY LEAMKEDLAE WLRDLYGLDI DAANFLQVLE TGLVLCQHAN VVTDAALAFL AEAPAQAQKI PMPRVGVSCN GAAQPGTFQA RDNVSNFIQW CRKEMGIQEV LMFETEDLVL RKNVKNVVLC LLELGRRAWR FGVAAPTLVQ LEEEIEEEVR RELALPPPDP SPPAPPRRQP CHFRNLDQMV QSLVSHCTCP VQFSMVKVSE GKYRVGDSNT LIFIRILRNH VMVRVGGGWD TLGHYLDKHD PCRCTSLSHK PGSFLKPPAP PVQHEVRVQD GPSQTQPTMT ISRSQSPPPP VDWKTYTSSD RRLRPPTPSS PRPRRERGAG TGASREMAPF LRCQERSLIP SWRQPTAGDS PPSPQSSSTQ KGRDPQCTSS GKREERYPPE LPRGRIPTSW VHEETDSWGT DAGNPTPQRL RAIEATTKGI SARGPSPLPR SFGPAECLGL RLPLRDEAKG AFFQFREPES VRSPTPVQGL TKIPIRLPPA RPPTPGRSFP GATSGSPRTE LGRDPIPLRA VTVDLAGSTH GDCSVEVRQE DQQLDIQVMA EARESWDLGL QEQEGRYTPL PLGGNKEQAI YCSLEEEILG NMKLLEVRSA CPQGTRSGVI PRSGVYIPRL AGQWPEPGGP YDKAIQELAQ GSPSLLKVDL EAWKAAPTGS PKPAVTPGPG SLKGKLGARQ SGPRTKASLS AKGTHMRKVP PQGGQDCSAS TVSASPEAPT PSPLDPNSDK AKACLSKGRR TLRKPKRVPS IYKLKLRPRI RPRRDHRPEK QPSRIPRPLA YVFLGPARQP PKDRLLRAVL GSKGGEASRV DGASVGEEEE EGKEEKEPAA PLESSPQPPE GLQPHWLNQA PLPPEEESWV

          Target Information

          Seems to be involved in the cross-linking of microtubules and microfilaments.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GAS2-like protein 2; GAS2-related protein on chromosome 17; Growth arrest-specific protein 2-like 2

          View more View less

          Gene Aliases: CILD41; GAR17; GAS2L2

          View more View less

          UniProt ID: (Human) Q8NHY3

          View more View less

          Entrez Gene ID: (Human) 246176

          View more View less

          Function(s)
          G-protein alpha-subunit binding protein binding microtubule binding cytoskeletal adaptor activity actin filament binding
          Process(es)
          microtubule bundle formation negative regulation of microtubule depolymerization regulation of microtubule polymerization or depolymerization positive regulation of G-protein coupled receptor protein signaling pathway actin crosslink formation regulation of cilium beat frequency involved in ciliary motility protein localization to microtubule plus-end
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0