Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SVSLSLKGS TETRSLPGET QSQGSLPLTW SFQEGVQLPG SYSGHLIANT PQQNSLNDCF SFSKSYGLGG ITDLTDQTST VDPMKLYEKR KLSSSRTYSS GDLLPPSASG VYSDHGDLQA WSKNAALGRN HLADNCYSNY PFPLTSWPCS FSPSQNSSEP FYQQLPLEPP AAKTGCPPLW PNPAGNLYEE KVHVDFNSYV QSPAYHSPQE DPFLFTYASH PHQQYSLPSK SSKWDFEEEM TYLGLDHCNN DMLLNLCPLR (178-436 aa encoded by BC096288)
This gene encodes a DNA-binding protein with a gcm-motif (glial cell missing motif). The encoded protein is a homolog of the Drosophila glial cells missing gene (gcm). This protein binds to the GCM-motif (A/G)CCCGCAT, a novel sequence among known targets of DNA-binding proteins. The N-terminal DNA-binding domain confers the unique DNA-binding activity of this protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCM motif protein 1; glial cells missing homolog 1; glial cells missing transcription factor 1; hGCMa
Gene Aliases: GCMA; hGCMa
UniProt ID: (Human) Q9NP62
Entrez Gene ID: (Human) 8521
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support