Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is predicted to react with rat based on sequence homology.
This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta1,6-N-acetylglucosaminyltransferase-M; beta1,6GlcNAc-transferase; C2GnT-M; C2GnT-mucin type; core 2 beta-1,6-N-acetylglucosaminyltransferase II; core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; Core2-GlcNAcT-II; glycosyltransferase; glycosyltransferase; GnT-M; hC2GnT-M; mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase; unnamed protein product
Gene Aliases: 2010013H22Rik; 2210021I22Rik; 2210401J11Rik; C2/4GnT; C24GNT; C2GNT2; C2GNTM; GNTM
UniProt ID: (Human) O95395, (Mouse) Q5JCT0
Entrez Gene ID: (Human) 9245, (Mouse) 72077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support