Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MCGIFAYLN YHVPRTRREI LETLIKGLQR LEYRGYDSAG VGFDGGNDKD WEANACKIQL IKKKGKVKAL DEEVHKQQDM DLDIEFDVHL GIAHTRWATH GEPSPVNSHP QRSDKNNEFI VIHNGIITNY KDLKKFLESK GYDFESETDT ETIAKLVKYM YDNRESQDTS FTTLVERVIQ QLEGAFALVF KSVHFPGQAV GTRRGSPLLI GVRSEHKLST DHIPILYRTG KDKKGSCNLS RVDSTTCLFP VEEKAVEYYF ASDASAVIEH TNRVIFLEDD DVAAVVDGRL SIHRIKRTAG DHPGRAVQTL QMELQQIMKG NFSSFMQKEI FEQPESVVNT MRGRVNFDDY T (1-350 aa encoded by BC045641)
This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-6-phosphate amidotransferase 1; GFAT 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine:fructose-6-phosphate amidotransferase 1; hexosephosphate aminotransferase 1; unnamed protein product
Gene Aliases: 2810423A18Rik; CMS12; CMSTA1; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; GFPT1L; MSLG
UniProt ID: (Human) Q06210, (Mouse) P47856, (Rat) P82808
Entrez Gene ID: (Human) 2673, (Mouse) 14583, (Rat) 297417
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support