Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • GGT1 Antibodies

          Invitrogen

          GGT1 Polyclonal Antibody

          View all (19) GGT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GGT1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          GGT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GGT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GGT1 Antibody (PA5-120579) in IHC (P)

          Immunohistochemistry analysis of GGT1 in paraffin-embedded human breast cancer using. Samples were incubated with GGT1 Polyclonal antibody (Product # PA5-120579) using a dilution of 1:100 (40x lens). Perform microwave antigen retrieval with 10 mM PBS buffer pH 7.2 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GGT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GGT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GGT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GGT1 Antibody in Western Blot (WB)
          GGT1 Polyclonal Antibody

          Product Details

          PA5-120579

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.64 mg/mL

          Amount

          64 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2914151

          Product Specific Information

          Positive test controls include: Jurkat, Mouse kidney. The target is usually found in the following locations: Cell membrane, Single-pass type II membrane protein.

          Immunogen sequence: KPEFYTPDDG GTAHLSVVAE DGSAVSATST INLYFGSKVR SPVSGILFNN EMDDFSSPSI TNEFGVPPSP ANFIQPGKQP LSSMCPTIMV GQDGQVRMVV GAAGGTQITT ATALAIIYNL WFGYDVKRAV EEPRLHNQLL PNVTTVERNI DQAVTAALET RHHHTQIAST FIAVVQAIVR TAGGWAAASD SRKGGEPAGY

          Target Information

          Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD224; gamma glutamyl-transferase 1; gamma-glutamyl transpeptidase; gamma-glutamyl transpeptidase (EC 2.3.2.2); Gamma-glutamyltransferase 1; gamma-glutamyltransferase 1 (EC 2.3.2.2); Gamma-glutamyltranspeptidase 1; GGT 1; GGT1; glutathione hydrolase 1; Glutathione hydrolase 1 proenzyme; Leukotriene-C4 hydrolase; testicular tissue protein Li 73

          View more View less

          Gene Aliases: CD224; D22S672; D22S732; dwg; GGLUT; GGT; GGT 1; GGT-1; GGT1; GGTD; Ggtp; GTG

          View more View less

          UniProt ID: (Human) P19440, (Mouse) Q60928, (Rat) P07314

          View more View less

          Entrez Gene ID: (Human) 2678, (Mouse) 14598, (Rat) 116568

          View more View less

          Function(s)
          peptidyltransferase activity leukotriene-C(4) hydrolase protein binding peptidase activity transferase activity transferase activity, transferring acyl groups hydrolase activity glutathione hydrolase activity leukotriene C4 gamma-glutamyl transferase activity gamma-glutamyltransferase activity protease
          Process(es)
          regulation of immune system process proteolysis cellular amino acid metabolic process glutamate metabolic process lipid metabolic process fatty acid metabolic process leukotriene metabolic process glutathione metabolic process glutathione biosynthetic process glutathione catabolic process spermatogenesis cysteine biosynthetic process peptide modification zymogen activation regulation of inflammatory response leukotriene D4 biosynthetic process aging leukotriene biosynthetic process response to estradiol response to lipopolysaccharide cellular response to oxidative stress response to tumor necrosis factor
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline