Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KAYPEVLAQ GRTARVTLTS PWPRPLPWPG NTLGQVGTPG TKDPRALLLD ALRSPGLRAL EAGTAVELLD VFLGLETDGE ELAGAIAAGN PGAPLRERAA ELREALEQGP RGLALRLWPK LQVVVTLDAG GQAEAVAALG ALWCQGLAFF SPAYAASGGV LGLNLQPEQP HGLYLLPPGA PFIELLPVKE GTQEEAASTL LLAEAQQGKE YELVLTDRAS LTRCRLGDVV RVVGAYNQCP VVRFICRLDQ TLSVRGEDIG EDLFSEALGR AVGQWAGAKL LDHGCVESSI L (140-429 aa encoded by BC022784)
GHDC, also known as LGP1, is a GH3 domain-containing protein that is highly expressed in normal mammary tissues and mammary tumors. GHDC is primarily located in the nuclear envelope. In mouse, its gene is located on chromosome 11 adjacent to the STAT3/5 locus. Little is known of the function of this protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D11lgp1; DL11LGP1; GHDC1; homolog of mouse LGP1; unnamed protein product
Gene Aliases: D11LGP1; D11Lgp1e; LGP1
UniProt ID: (Human) Q8N2G8, (Mouse) Q99J23
Entrez Gene ID: (Human) 84514, (Mouse) 80860
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support