Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GLI3 Antibodies

          Invitrogen

          GLI3 Polyclonal Antibody

          Advanced Verification
          1 Published Figure
          3 References
          View all (14) GLI3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GLI3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          • Published Figures (1)
          • Advanced Verification (1)
          GLI3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GLI3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GLI3 Antibody (PA5-28029) in ICC/IF

          Immunocytochemistry analysis of GLI3 in paraformaldehyde-fixed HeLa. Samples were incubated with GLI3 polyclonal antibody (Product # PA5-28029) using a dilution of 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GLI3 Antibody in Immunocytochemistry (ICC/IF)
          GLI3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GLI3 Antibody in Western Blot (WB)
          GLI3 Antibody in Western Blot (WB)
          GLI3 Antibody in Western Blot (WB)
          GLI3 Antibody in Western Blot (WB)
          GLI3 Antibody in Western Blot (WB)
          GLI3 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          GLI3 Antibody

          Product Details

          PA5-28029

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human GLI3. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.43 mg/mL

          Amount

          43 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 1% BSA, 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2545505

          Product Specific Information

          Recommended positive controls: Jurkat, mouse ESC, PC-12, Rat2.

          Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (87%), Chicken (91%), Chimpanzee (100%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          This gene encodes a protein which belongs to the C2H2-type zinc finger proteins subclass of the Gli family. They are characterized as DNA-binding transcription factors and are mediators of Sonic hedgehog (Shh) signaling. The protein encoded by this gene localizes in the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. Mutations in this gene have been associated with several diseases, including Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, and postaxial polydactyly types A1 and B.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: anterior digit pattern deformity; brachyphalangy; extra toes; GLI-Kruppel family member GLI3; GLI3 form of 190 kDa; GLI3 full length protein; GLI3 full-length protein; GLI3-190; GLI3FL; glioma-associated oncogene family zinc finger 3; oncogene GLI3; polydactyly Nagoya; Transcriptional activator GLI3; unnamed protein product; zinc finger protein GLI3

          View more View less

          Gene Aliases: ACLS; add; AI854843; AU023367; Bph; GCPS; GLI3; GLI3-190; GLI3FL; PAP-A; PAPA; PAPA1; PAPB; Pdn; PHS; PPDIV; Xt

          View more View less

          UniProt ID: (Human) P10071, (Mouse) Q61602

          View more View less

          Entrez Gene ID: (Human) 2737, (Rat) 140588, (Mouse) 14634

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding chromatin binding double-stranded DNA binding transcription factor activity, sequence-specific DNA binding protein binding beta-catenin binding zinc ion binding histone acetyltransferase binding mediator complex binding histone deacetylase binding sequence-specific DNA binding metal ion binding sequence-specific double-stranded DNA binding nucleic acid binding DNA-binding transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter osteoblast differentiation metanephros development branching involved in ureteric bud morphogenesis in utero embryonic development kidney development positive regulation of neuroblast proliferation chondrocyte differentiation regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter protein import into nucleus apoptotic process smoothened signaling pathway pattern specification process neuroblast proliferation axon guidance central nervous system development brain development hindgut morphogenesis heart development cell proliferation negative regulation of cell proliferation regulation of smoothened signaling pathway anatomical structure morphogenesis anterior/posterior pattern specification dorsal/ventral pattern formation proximal/distal pattern formation gene expression regulation of gene expression protein processing spinal cord dorsal/ventral patterning spinal cord motor neuron differentiation telencephalon development pallium development subpallium development optic nerve morphogenesis hippocampus development smoothened signaling pathway involved in ventral spinal cord interneuron specification smoothened signaling pathway involved in spinal cord motor neuron cell fate specification forebrain dorsal/ventral pattern formation cerebral cortex radial glia guided migration layer formation in cerebral cortex forebrain radial glial cell differentiation neural tube development central nervous system neuron differentiation lateral ganglionic eminence cell proliferation melanocyte differentiation lung development embryonic limb morphogenesis mammary gland development forebrain development negative regulation of chondrocyte differentiation positive regulation of chondrocyte differentiation T cell differentiation in thymus limb morphogenesis tube development regulation of cell proliferation positive regulation of protein import into nucleus odontogenesis of dentin-containing tooth embryonic digit morphogenesis regulation of apoptotic process camera-type eye development negative regulation of apoptotic process nose morphogenesis tongue development negative thymic T cell selection regulation of cell differentiation negative regulation of neuron differentiation positive regulation of osteoblast differentiation negative regulation of smoothened signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter alpha-beta T cell differentiation positive regulation of alpha-beta T cell differentiation negative regulation of alpha-beta T cell differentiation embryonic digestive tract morphogenesis embryonic digestive tract development developmental growth camera-type eye morphogenesis embryonic morphogenesis anatomical structure formation involved in morphogenesis neuron fate commitment embryonic neurocranium morphogenesis embryonic skeletal system morphogenesis oligodendrocyte differentiation branching morphogenesis of an epithelial tube inner ear development palate development limb development frontal suture morphogenesis lambdoid suture morphogenesis sagittal suture morphogenesis mammary gland specification smoothened signaling pathway involved in dorsal/ventral neural tube patterning artery development anterior semicircular canal development lateral semicircular canal development cell differentiation involved in kidney development thymocyte apoptotic process vocalization behavior stem cell proliferation negative regulation of canonical Wnt signaling pathway larynx morphogenesis regulation of bone development negative regulation of stem cell proliferation transcription, DNA-templated negative regulation of cell differentiation anatomical structure development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-zb4k5:80/100.66.74.142:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline