Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSESLVVCD VAEDLVEKLR KFRFRKETNN AAIIMKIDKD KRLVVLDEEL EGISPDELKD ELPERQPRFI VYSYKYQHDD GRVSYPLCFI FSSPVGCKPE QQMMYAGSKN KLVQTAELTK VFEIRNTEDL TEEWLREKLG FFTNVNFCVS KVFMY (1-154 aa encoded by BC005359)
This protein, which is phosphorylated following phorbol ester stimulation, is important for the nervous system. It causes brain cell differentiation, stimulates neural regeneration and inhibits tumor cell proliferation. Overexpression of GMF in astrocytes has been shown to enhance brain-derived neurotrohic factor (BDNF) production. GMF expression is increased by exercise, and the protein is crucial for exercise-induction of BDNF. Through BDNF production, GMF appears to play a role in neuroprotection. In thymoma, T-cell development is maintained by GMF; being produced by the tumor cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA for thyroid hormone receptor binding site (276bp); GMF-beta; GMFB; unnamed protein product
Gene Aliases: 3110001H22Rik; 3110001O16Rik; D14Ertd630e; GMF
UniProt ID: (Human) P60983, (Rat) Q63228, (Mouse) Q9CQI3
Entrez Gene ID: (Human) 2764, (Rat) 81661, (Mouse) 63985
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support