Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • GPR56 Antibodies

          Abnova

          GPR56 Polyclonal Antibody, MaxPab™

          View all (19) GPR56 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GPR56 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          GPR56 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GPR56 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GPR56 Antibody (H00009289-D01P) in WB

          Western Blot analysis of GPR56 expression in transfected 293T cell line by GPR56 MaxPab polyclonal antibody. Lane 1: GPR56 transfected lysate(77.70 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GPR56 Antibody in Western Blot (WB)

          Product Details

          H00009289-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GPR56 (AAH08770.1, 1 a.a. approximately 693 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MTPQSLLQTT LFLLSLLFLV QGAHGRGHRE DFRFCSQRNQ THRSSLHYKP TPDLRISIEN SEEALTVHAP FPAAHPASRS FPDPRGLYHF CLYWNRHAGR LHLLYGKRDF LLSDKASSLL CFQHQEESLA QGPPLLATSV TSWWSPQNIS LPSAASFTFS FHSPPHTAAH NASVDMCELK RDLQLLSQFL KHPQKASRRP SAAPASQQLQ SLESKLTSVR FMGDMVSFEE DRINATVWKL QPTAGLQDLH IHSRQEEEQS EIMEYSVLLP RTLFQRTKGR SGEAEKRLLL VDFSSQALFQ DKNSSHVLGE KVLGIVVQNT KVANLTEPVV LTFQHQLQPK NVTLQCVFWV EDPTLSSPGH WSSAGCETVR RETQTSCFCN HLTYFAVLMV SSVEVDAVHK HYLSLLSYVG CVVSALACLV TIAAYLCSRV PLPCRRKPRD YTIKVHMNLL LAVFLLDTSF LLSEPVALTG SEAGCRASAI FLHFSLLTCL SWMGLEGYNL YRLVVEVFGT YVPGYLLKLS AMGWGFPIFL VTLVALVDVD NYGPIILAVH RTPEGVIYPS MCWIRDSLVS YITNLGLFSL VFLFNMAMLA TMVVQILRLR PHTQKWSHVL TLLGLSLVLG LPWALIFFSF ASGTFQLVVL YLFSIITSFQ GFLIFIWYWS MRLQARGGPS PLKSNSDSAR LPISSGSTSS SRI

          Target Information

          GPR56 is a member of the G protein-coupled receptor family. The protein contains 7 transmembrane domains and a mucin-like domain in the N-terminal region. The gene is implicated in the regulation of brain cortical patterning. The protein binds specifically to transglutaminase 2 in the extracellular space. Expression of this gene is downregulated in melanoma cell lines, and overexpression of this gene can suppress tumor growth and metastasis. Mutations in this gene result in bilateral frontoparietal polymicrogyria. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 7-transmembrane protein with no EGF-like N-terminal domains-1; Adhesion G-protein coupled receptor G1; Cyt28; DKFZp781L1398; G protein-coupled receptor 56; G-protein coupled receptor 56; SLR32; testicular tissue protein Li 77; unnamed protein product

          View more View less

          Gene Aliases: ADGRG1; BFPP; BPPR; CDCBM14B; CDCBM15A; GPR56; TM7LN4; TM7XN1; UNQ540/PRO1083

          View more View less

          UniProt ID: (Human) Q9Y653

          View more View less

          Entrez Gene ID: (Human) 9289

          View more View less

          Function(s)
          transmembrane signaling receptor activity G-protein coupled receptor activity protein binding collagen binding heparin binding extracellular matrix binding
          Process(es)
          angiogenesis cell adhesion signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway Rho protein signal transduction cell-cell signaling nervous system development brain development negative regulation of cell proliferation positive regulation of signal transduction cell migration cerebral cortex regionalization cerebral cortex radial glia guided migration layer formation in cerebral cortex cell differentiation positive regulation of Rho protein signal transduction tube development regulation of cell proliferation positive regulation of cell adhesion anatomical structure formation involved in morphogenesis neural precursor cell proliferation hematopoietic stem cell homeostasis seminiferous tubule development regulation of platelet aggregation negative regulation of ferroptosis Rho-activating G protein-coupled receptor signaling pathway positive regulation of vascular endothelial growth factor signaling pathway positive regulation of neural precursor cell proliferation negative regulation of neuron migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline