PA1-24572 detects G Protein-Coupled Receptor GPR88 in Human samples.
PA1-24572 has been successfully used in Immunohistochemistry (paraffin) procedures.
PA1-24572 immunogen corresponds to Synthetic peptide - KLH conjugated. Sequence identity/similarity to Rat: 94/94%.
GPR88 is an Orphan-A GPCR with an unknown ligand. GPR88 expression has been documented almost exclusively in the striatum of the brain and not in any peripheral tissues. ESTs have been isolated from diseased brain, ear, and kidney libraries.