PA1-25534 detects G Protein-Coupled Receptor 5C from human samples.
PA1-25534 has been successfully used in immunohistochemistry (paraffin) procedures.
The PA1-25534 immunogen is a synthetic peptide corresponding to a region of human G Protein-Coupled Receptor 5C.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This polyclonal antibody was raised in Rabbit against GPRC5C. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPRC5C. BLAST analysis of the peptide immunogen showed no homology with other human proteins.