Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
16757-1-AP antibody recognizes two different isoforms of GPT2 protein.
Immunogen sequence: VPADPDNIY LTTGASDGIS TILKILVSGG GKSRTGVMIP IPQYPLYSAV ISELDAIQVN YYLDEENCWA LNVNELRRAV QEAKDHCDPK VLCIINPGNP TGQVQSRKCI EDVIHFAWEE KLFLLADEVY QDNVYSPDCR FHSFKKVLYE MGPEYSSNVE LASFHSTSKG YMGECGYRGG YMEVINLHPE IKGQLVKLLS VRLCPPVSGQ AAMDIVVNPP VAGEESFEQF SREKESVLGN LAKKAKLTED LFNQVPGIHC NPLQGAMYAF PRIFIPAKAV EAAQAHQMAP DMFYCMKLLE ETGICVVPGS GFGQREGTYH FRMTILPPVE KLKTVLQKVK DFHINFLEKY A (174-523 aa encoded by BC062555)
GPT and GPT2 (EC 2.6.1.2), also known as alanine transaminases, are pyridoxal enzymes that catalyze the reversible transamination between alanine and 2-oxoglutarate to form pyruvate and glutamate. By mediating the conversion of these 4 major intermediate metabolites, these transaminases have roles in gluconeogenesis and in amino acid metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALT2; glutamate pyruvate transaminase 2; glutamic pyruvate transaminase (alanine aminotransferase) 2; glutamic pyruvate transaminase 2; glutamic--alanine transaminase 2; glutamic--pyruvic transaminase 2; liver regeneration-related protein LRRG168; unnamed protein product
Gene Aliases: 4631422C05Rik; ALT2; Cc2-5; GPT 2; MRT49; NEDSPM
UniProt ID: (Human) Q8TD30, (Mouse) Q8BGT5
Entrez Gene ID: (Human) 84706, (Mouse) 108682, (Rat) 307759
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support