Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is specific to GSR.
Immunogen sequence: ISTNCTEEL ENAGVEVLKF SQVKEVKKTL SGLEVSMVTA VPGRLPVMTM IPDVDCLLWA IGRVPNTKDL SLNKLGIQTD DKGHIIVDEF QNTNVKGIYA VGDVCGKALL TPVAIAAGRK LAHRLFEYKE DSKLDYNNIP TVVFSHPPIG TVGLTEDEAI HKYGIENVKT YSTSFTPMYH AVTKRKTKCV MKMVCANKEE KVVGIHMQGL GCDEMLQGFA VAVKMGATKA DFDNTVAIHP TSSEELVTLR (274-522 aa encoded by BC069244)
This gene encodes a member of the class-I pyridine nucleotide-disulfide oxidoreductase family. This enzyme is a homodimeric flavoprotein. It is a central enzyme of cellular antioxidant defense, and reduces oxidized glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. Rare mutations in this gene result in hereditary glutathione reductase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; glutathione reductase 1; glutathione reductase, mitochondrial; glutathione S-reductase; GR; GRase; unnamed protein product
Gene Aliases: CNSHA10; D8Ertd238e; GR; Gr-1; Gr1; GSRD; HEL-75; HEL-S-122m
UniProt ID: (Human) P00390, (Mouse) P47791
Entrez Gene ID: (Human) 2936, (Mouse) 14782, (Rat) 116686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support