Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAEKPKLHY FNARGRMEST RWLLAAAGVE FEEKFIKSAE DLDKLRNDGY LMFQQVPMVE IDGMKLVQTR AILNYIASKY NLYGKDIKER ALIDMYIEGI ADLGEMILLL PVCPPEEKDA KLALIKEKIK NRYFPAFEKV LKSHGQDYLV GNKLSRADIH LVELLYYVEE LDSSLISSFP LLKALKTRIS NLPTVKKFLQ PGSPRKPPMD EKSLEEARKI FRF (1-222 aa encoded by BC053578)
This gene encodes a member of a family of enzymes that function to add glutathione to target electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins, and products of oxidative stress. This action is an important step in detoxification of these compounds. This subfamily of enzymes has a particular role in protecting cells from reactive oxygen species and the products of peroxidation. Polymorphisms in this gene influence the ability of individuals to metabolize different drugs. This gene is located in a cluster of similar genes and pseudogenes on chromosome 6. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 13-hydroperoxyoctadecadienoate peroxidase; androst-5-ene-3,17-dione isomerase; class alpha; detoxification enzyme; glutathione S-alkyltransferase A1; glutathione S-aryltransferase A1; glutathione S-transferase (GST, EC 2.5.1.18); glutathione S-transferase 2; glutathione S-transferase A1; glutathione S-transferase A3 subunit; glutathione S-transferase A5; glutathione S-transferase Ha subunit 1; glutathione S-transferase Ha subunit 1 (EC 2.5.1.18); glutathione S-transferase Ya; glutathione S-transferase Ya1; glutathione S-transferase Yc subunit; glutathione S-transferase Yc1 subunit; Glutathione-S-transferase alpha type (Ya); gluthione S-transferase subunit 1 (GST,EC 2.5.1.18); GST; GST class-alpha member 1; GST HA subunit 1; GSTA1; RP11-392A23.1; S-(hydroxyalkyl)glutathione lyase A1; testicular tissue protein Li 80; unnamed protein product
Gene Aliases: GST 2-2; GST A3-3; GST AA; GST Yc1; GST-epsilon; GST2; Gst2-1; GSTA1-1; Gsta3; Gsta5; GTH1; Yc1
UniProt ID: (Human) P08263, (Rat) P04904, (Mouse) P13745
Entrez Gene ID: (Human) 2938, (Rat) 24421, (Mouse) 14857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support