Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • GSTT1 Antibodies

          Proteintech

          GSTT1 Polyclonal Antibody

          9 References
          View all (8) GSTT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GSTT1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (28)
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 28

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GSTT1 Antibody (15838-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human kidney tissue slide using 15838-1-AP (GSTT1 Antibody) at dilution of 1:50 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTT1 Antibody in Western Blot (WB)
          GSTT1 Antibody in Western Blot (WB)
          GSTT1 Antibody in Western Blot (WB)
          GSTT1 Antibody in Western Blot (WB)
          GSTT1 Antibody in Western Blot (WB)
          GSTT1 Antibody in Western Blot (WB)
          GSTT1 Polyclonal Antibody

          Product Details

          15838-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Hamster, Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GSTT1 Fusion Protein Ag8588 (1-240 aa encoded by BC007065)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGLELYLDL LSQPCRAVYI FAKKNDIPFE LRIVDLIKGQ HLSDACAQVN PLKKVPALKD GDFTLTESVA ILLYLTRKYK VPDYWYPQDL QARARVDEYL AWQHTTLRRS CLRALWHKVM FPVFLGEPVS PQTLAATLAE LDVTLQLLED KFLQNKAFLT GPHISLADLV AITELMHPVG AGCQVFEGRP KLATWRQRVE AAVGEDLFQE AHEVILKAKD FPPADPTIKQ KLMPWVLAMI R (1-240 aa encoded by BC007065)

          Target Information

          Glutathione S-transferase (GST) theta 1 (GSTT1) is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Glutathione S-transferase 1 (theta); Glutathione S-transferase 5; Glutathione S-transferase theta-1; Glutathione transferase T1-1; GST 5-5; GST class-theta-1; GSTT1

          View more View less

          Gene Aliases: AI255817; GSTT1; Gstt1-1; GSTYRS

          View more View less

          UniProt ID: (Human) P30711, (Rat) Q01579, (Mouse) Q64471

          View more View less

          Entrez Gene ID: (Human) 2952, (Rat) 25260, (Mouse) 14871

          View more View less

          Function(s)
          glutathione transferase activity glutathione peroxidase activity protein binding transferase activity alkylhalidase activity
          Process(es)
          glutathione metabolic process cellular oxidant detoxification DNA modification response to salicylic acid response to selenium ion response to organic cyclic compound dichloromethane metabolic process response to vitamin E response to drug oxidation-reduction process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0