Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MATSSEEVLL IVKKVRQKKQ DGALYLMAER IAWAPEGKDR FTISHMYADI KCQKISPEGK AKIQLQLVLH AGDTTNFHFS NESTAVKERD AVKDLLQQLL PKFKRKANKE LEEKNRMLQE DPVLFQLYKD LVVSQVISAE EFWANRLNVN ATDSSSTSNH KQDVGISAAF LADVRPQTDG CNGLRYNLTS DIIESIFRTY PAVKMKYAEN VPHNMTEKEF WTRFFQSHYF HRDRLNTGSK DLFAECAKID EKGLKTMVSL GVKNPLLDLT ALEDKPLDEG YGISSVPSAS NSKSIKENSN AAIIKRFNHH SAMVLAAGLR KQEAQNEQTS EPSNMDGNSG DADCFQPAVK RAKLQESIEY EDLGKNNSVK TIALNLKKSD RYYHGPTPIQ SLQYATSQDI INSFQSIRQE MEAYTPKLTQ VLSSSAASST ITALSPGGAL MQGGTQQAIN QMVPNDIQSE LKHLYVAVGE LLRHFWSCFP VNTPFLEEKV VKMKSNLERF QVTKLCPFQE KIRRQYLSTN LVSHIEEMLQ TAYNKLHTWQ SRRLMKKT
GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa) is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA. When complexed to CAK, GTF2H1 is involved in RNA transcription by RNA polymerase II.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic transcription factor 2 62 kDa subunit; BTF 2; BTF2 p62; BTF2. TFIIH; General transcription factor IIH polypeptide 1; General transcription factor IIH subunit 1; general transcription factor IIH, polypeptide 1, 62kDa; TF2H1; TFIIH basal transcription factor complex p62 subunit
Gene Aliases: BTF2; GTF2H1; P62; TFB1; TFIIH
UniProt ID: (Human) P32780
Entrez Gene ID: (Human) 2965
Molecular Function: RNA metabolism protein nucleic acid metabolism protein general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support