Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 4
CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCK(B) receptor; CCK-B receptor; CCK-BR; CCK2 receptor; CCK2-R; CCKB; Cckb receptor; CCKB-R; Cholecystokinin-2 receptor; Cholecystokinin-B; gastrin receptor; Gastrin/cholecystokinin type B receptor; HGNC:1571
Gene Aliases: CCK-B; CCK-BR; CCK2-R; CCK2R; CCKBR; CCKR-2; CCKRB; Cholrec; GASR
UniProt ID: (Human) P32239, (Rat) P30553, (Mouse) P56481
Entrez Gene ID: (Human) 887, (Rat) 25706, (Mouse) 12426
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support