Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • GluR6 Antibodies

          Invitrogen

          GluR6 Polyclonal Antibody

          View all (17) GluR6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GluR6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          GluR6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GluR6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GluR6 Antibody (PA5-96001) in WB

          Western blot analysis of GluR6 in extracts of various cell lines. Samples were incubated with GluR6 Polyclonal antibody (Product # PA5-96001) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GluR6 Antibody in Western Blot (WB)
          GluR6 Polyclonal Antibody

          Product Details

          PA5-96001

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 30-300 of human GRIK2 (NP_0687751)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.54 mg/mL

          Amount

          54 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2807803

          Product Specific Information

          Immunogen sequence: QGTTHVLRFG GIFEYVESGP MGAEELAFRF AVNTINRNRT LLPNTTLTYD TQKINLYDSF EASKKACDQL SLGVAAIFGP SHSSSANAVQ SICNALGVPH IQTRWKHQVS DNKDSFYVSL YPDFSSLSRA ILDLVQFFKW KTVTVVYDDS TGLIRLQELI KAPSRYNLRL KIRQLPADTK DAKPLLKEMK RGKEFHVIFD CSHEMAAGIL KQALAMGMMT EYYHYIFTTL DLFALDVEPY RYSGVNMTGF RILNTENTQV SSIIEKWSME R; Positive Samples: SW620, U-251MG; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse

          Target Information

          Glutamate receptors are the primary excitatory neurotransmitter receptors in the mammalian brain and are activated during various normal neurophysiological processes. The kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels, includes this gene product. The subunit encoded by this gene is edited at multiple sites within the first and second transmembrane domains, which is thought to alter the structure and function of the receptor complex. Furthermore, different isoforms of this gene have been described due to alternatively spliced transcript variants. Mutations in this gene have been linked to autosomal recessive cognitive disability. GRIK2-associated diseases include Neurodevelopmental Disorder with Impaired Language and Ataxia and with or without Seizures and Intellectual Developmental Disorder, Autosomal Recessive 6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bA487F5.1; EAA4; Excitatory amino acid receptor 4; GLR 6; GluK2; GluK2(alt_5'UTR); GLUR 6; GluR beta-2; GluR-6; glutamate receptor; Glutamate receptor 6; Glutamate receptor beta-2; glutamate receptor form A; glutamate receptor form B; glutamate receptor form C; glutamate receptor form D; glutamate receptor form E; Glutamate receptor ionotropic, kainate 2; glutamate receptor, ionotropic kainate 2; GRIK 2; GRIK2 protein; kainate 2; putative NMDtranscript(cassette_171nt); unnamed protein product

          View more View less

          Gene Aliases: AW124492; EAA4; GLR6; GluK2; GLUK6; Glur-6; GLUR6; Glurbeta2; GRIK2; MRT6; NEDLAS

          View more View less

          UniProt ID: (Human) Q13002, (Mouse) P39087

          View more View less

          Entrez Gene ID: (Human) 2898, (Mouse) 14806

          View more View less

          Function(s)
          SNARE binding ionotropic glutamate receptor activity ion channel activity extracellular-glutamate-gated ion channel activity glutamate receptor activity ligand-gated ion channel activity kainate selective glutamate receptor activity glutamate-gated calcium ion channel activity PDZ domain binding ubiquitin conjugating enzyme binding ubiquitin protein ligase binding signaling receptor activity identical protein binding scaffold protein binding ligand-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential receptor activity protein binding protein homodimerization activity transmembrane signal receptor
          Process(es)
          behavioral fear response ion transport cellular calcium ion homeostasis glutamate receptor signaling pathway chemical synaptic transmission neuronal action potential calcium-mediated signaling ion transmembrane transport ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic regulation of membrane potential receptor clustering negative regulation of neuron apoptotic process positive regulation of neuron apoptotic process regulation of JNK cascade regulation of long-term neuronal synaptic plasticity regulation of short-term neuronal synaptic plasticity modulation of synaptic transmission positive regulation of synaptic transmission neuron apoptotic process negative regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential inhibitory postsynaptic potential modulation of excitatory postsynaptic potential presynaptic modulation of chemical synaptic transmission regulation of presynaptic membrane potential detection of cold stimulus involved in thermoception transport intracellular protein transport signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline