Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Secondary Antibodies ›
          • Goat anti-Rabbit IgG Fc Secondary Antibodies

          Invitrogen

          Goat anti-Rabbit IgG Fc Superclonal™ Secondary Antibody, Alexa Fluor™ 488

          Advanced Verification
          View all (36) Goat anti-Rabbit IgG Fc antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Goat anti-Rabbit IgG Fc Superclonal™ Secondary Antibody, Alexa Fluor™ 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          Rabbit IgG Fc Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rabbit IgG Fc Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rabbit IgG Fc Secondary Antibody (SA5-10378-AF488) in ICC/IF

          Immunofluorescence analysis of Goat anti-Rabbit IgG Fc Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 488 (Product # SA5-10378-AF488) was performed on fixed and permeabilized HeLa cells blocked with SuperBlock™ Blocking Buffer (Product # 37515) at room temperature and stained with H3K4me3 Recombinant Rabbit Monoclonal Antibody (14H24L5) (Product # 703849, 1:250) in SuperBlock™ Blocking Buffer (Pr... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rabbit IgG Fc Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG Fc Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rabbit IgG Fc Secondary Antibody

          Product Details

          SA5-10378-AF488

          Applications
          Tested Dilution

          Immunohistochemistry (Paraffin) (IHC (P))

          1:250

          Immunocytochemistry (ICC/IF)

          1:1,000
          Product Specifications

          Species Reactivity

          Rabbit

          Host/Isotype

          Goat / IgG

          Expression System

          Expi293

          Class

          Recombinant Superclonal

          Type

          Secondary Antibody

          Immunogen

          Purified Rabbit IgG

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Alexa Fluor 647 
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 647 
          • Alexa Fluor Plus 680 
          • AP
          • Biotin 
          • FITC
          • HRP
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          250 µg

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 6.3-7.3

          Contains

          0.008% Bromonitrodioxane, 0.008% Methylisothiazolone

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_3719773

          Target

          IgG Fc

          Antibody Form

          Whole Antibody

          Product Specific Information

          The Goat anti-Rabbit IgG Fc Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 488 is specific to Fc region of Rabbit IgG with moderate cross reactivity to Monkey IgG. It showed no cross-reactivity to any other immunoglobulins tested from other species.

          The sensitivity and specificity of each lot is confirmed using ELISA.

          Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency.

          Protein conjugates made with Alexa Fluor™ dyes produce fluorescence output that surpasses that of other spectrally similar fluorophore-labeled proteins. Alexa Fluor™ dyes are available in a wide selection of fluorescent colors and are more photostable than most other dyes. Photostability is a key characteristic of Alexa Fluor dye™ conjugates, allowing more time for image capture. In addition to their exceptional brightness and the wide selection of dyes across the spectrum, you'll get photostability you can rely on with Alexa Fluor™ dyes and dye conjugates.

          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Target Information

          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody SA5-10378-AF488 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline