Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Rabbit IgG (Heavy chain) Secondary Antibodies

          Invitrogen

          Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, HRP

          5 Published Figures
          View all (10) Goat anti-Rabbit IgG (Heavy chain) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, HRP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          • Published Figures (5)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rabbit IgG (Heavy chain) Secondary Antibody (A27036SAMPLE) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of A-431 (Lane1), COS-7 (Lane 2), HT-29 (Lane 3), Jurkat (Lane 4), Raji (lane 5), Ramos (Lane 6), Daudi (Lane 7) and HEL (Lane 8). The blots were probed with Anti-Profilin-1 Rabbit Polyclonal Antibody (Product # 720121, 1-2 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunoprecipitation (IP)
          Rabbit IgG (Heavy chain) Secondary Antibody in ELISA (ELISA)
          Rabbit IgG (Heavy chain) Secondary Antibody in Peptide array (ARRAY)
          Rabbit IgG (Heavy chain) Secondary Antibody in Peptide array (ARRAY)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)

          Product Details

          A27036SAMPLE

          Applications
          Tested Dilution

          Western Blot (WB)

          1:10,000-1:200,000

          ELISA (ELISA)

          0.05-1 µg/mL

          Immunoprecipitation (IP)

          1:1,000-1:5,000

          Peptide Array (Array)

          1:1,000-1:5,000
          Product Specifications

          Species Reactivity

          Rabbit

          Host/Isotype

          Goat / IgG

          Expression System

          Expi293

          Class

          Recombinant Superclonal

          Type

          Secondary Antibody

          Immunogen

          Recombinant full-length protein

          Conjugate

          HRP HRP HRP
          • Alexa Fluor 488 
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 790 
          • Alexa Fluor Plus 405 
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 647 
          • Biotin 
          • Unconjugated
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Gravity column chromatography

          Storage buffer

          PBS with HRP Stabilizer

          Contains

          proprietary preservative

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_2610665

          Target

          IgG (Heavy chain)

          Antibody Form

          Whole Antibody

          Product Specific Information

          The sensitivity and specificity of each lot is confirmed using ELISA.

          Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed.

          Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.

          Target Information

          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody A27036SAMPLE on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline