          • Primary Antibodies ›
          • CDR2 Antibodies
          Abnova
          CDR2 Monoclonal Antibody (4F5)
          View all (10) CDR2 antibodies
          Tech Support
          Cite CDR2 Monoclonal Antibody (4F5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          CDR2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CDR2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          CDR2 Antibody (H00001039-M01) in ICC/IF
          Immunofluorescence of monoclonal antibody to CDR2 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CDR2 Antibody in Immunocytochemistry (ICC/IF)
          CDR2 Antibody in Western Blot (WB)
          CDR2 Antibody in Western Blot (WB)
          CDR2 Antibody in ELISA (ELISA)
          CDR2 Monoclonal Antibody (4F5)
          Product Details
          H00001039-M01
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-5 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          10 µg/mL
          -
          ELISA (ELISA)
          0.03 ng/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4F5
          Immunogen
          CDR2 (NP_001793, 296 a.a. approximately 404 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          See Label
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Sequence of this protein is as follows: LTVPESHRKP LKRSSSETIL SSLAGSDIVK GHEETCIRRA KAVKQRGISL LHEVDTQYSA LKVKYEELLK KCQEEQDSLS HKAVQTSRAA AKDLTGVNAQ SEPVASGWE
          Target Information
          Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.
          Bioinformatics
          Protein Aliases: Cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; Major Yo paraneoplastic antigen; Paraneoplastic cerebellar degeneration-associated antigen; unnamed protein product; Yo paraneoplastic antigen
          View more View less
          Gene Aliases: CDR2; CDR62; PCD17; Yo
          View more View less
          UniProt ID: (Human) Q01850
          View more View less
          Entrez Gene ID: (Human) 1039
          View more View less
          Function(s)
          protein binding
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline