    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • KLHDC4 Antibodies
          Abnova
          KLHDC4 Monoclonal Antibody (4G11)
          View all (7) KLHDC4 antibodies
          Tech Support
          Cite KLHDC4 Monoclonal Antibody (4G11)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          KLHDC4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KLHDC4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          KLHDC4 Antibody (H00054758-M02) in ICC/IF
          Immunofluorescence of monoclonal antibody to KLHDC4 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KLHDC4 Antibody in Immunocytochemistry (ICC/IF)
          KLHDC4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KLHDC4 Antibody in Western Blot (WB)
          KLHDC4 Antibody in ELISA (ELISA)
          KLHDC4 Monoclonal Antibody (4G11)
          Product Details
          H00054758-M02
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-5 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1.5 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          10 µg/mL
          -
          ELISA (ELISA)
          0.3 ng/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4G11
          Immunogen
          KLHDC4 (AAH01044, 1 a.a. approximately 463 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          See Label
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Sequence of this protein is as follows: MGKKGKKEKK GRGAEKTAAK MEKKVSKRSR KEETFLYNEL YVYNTRKDTW TKVDIPSPPP RRCAHQAVVV PQGGGQLWVF GGEFASPNGE QFYHYKDLWV LHLATKTWEQ VKSTGGPSGR SGHRMVAWKR QLILFGGFHE STRDYIYYND VYAFNLDTFT WSKLSPSGTG PTPRSGCQMS VTPQGGIVVY GGYSKQRVKK DVDKGTRHSD MFLLKPEDGR EDKWVWTRMN PSGVKPTPRS GFSVAMAPNH QTLFFGGVCD EEEEESLSGE FFNDLYFYDA TRNRWFEGQL KGPKSEKKKR RRGRKEEPEG GSRPACGGAG TQGPVQLVKE VVAEDGTVVT IKQVLTAPGS AGQPRSEDED SLEEAGSPAP GPCPRSNAML AVKHGVLYVY GGMFEAGDRQ VTLSDLHCLD LHRMEAWKAL VEMDPETQEW LEETDSEEDS EEVEGAEGGV DDEDSGEESG AED
          Target Information
          Kelch proteins (and Kelch-like proteins) are proteins that have repeated kelch domains. Most Kelch proteins also have a BTB (defn) domain. Each kelch domain forms the blade of a propellar structure, with the repeated kelch domains forming a Beta-propellar. The structure of this Beta-propellar is known but the function is not characterised. KLHDC4 contains six Kelch repeats. There are three named isoforms.
          Bioinformatics
          Protein Aliases: Kelch domain-containing protein 4; unnamed protein product
          View more View less
          Gene Aliases: KLHDC4
          View more View less
          UniProt ID: (Human) Q8TBB5
          View more View less
          Entrez Gene ID: (Human) 54758
          View more View less
          Function(s)
          protein binding
          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          Korea flag icon
          Korea
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)
          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)
          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline