Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 9
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Keep as concentrated solution.
Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (89%), Pig (100%), Chicken (95%), Rhesus Monkey (100%), Bovine (100%).
Positive Control: 293T, A431, HeLa, HepG2, U87-MG, SK-N-SH, IMR32, SK-N-AS, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2, NB2a.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HD3; HDAC3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; protein deacetylase HDAC3; protein deacylase HDAC3; SMAP45; transcriptional corepressor; unnamed protein product
Gene Aliases: HD3; KDAC3; RPD3; RPD3-2; zgc:55927
UniProt ID: (Human) O15379, (Mouse) O88895, (Rat) Q6P6W3, (Zebrafish) Q803C3
Entrez Gene ID: (Human) 8841, (Mouse) 15183, (Rat) 84578, (Zebrafish) 393965
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support