Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FPDREENAV HSTEPVVQEN GDEAGEGREA KDCDPGSPRR CDIIIISGRK EKCEAAKEAL EALVPVTIEV EVPFDLHRYV IGQKGSGIRK MMDEFEVNIH VPAPELQSDI IAITGLAANL DRAKAGLLER VKELQAEQED RALRSFKLSV TVDPKYHPKI IGRKGAVITQ IRLEHDVNIQ FPDKDDGNQP QDQITITGYE KNTEAARDAI LRIVGELEQM VSEDVPLDHR VHARIIGARG KAIRKIMDEF KVDIRFPQSG APDPNCVTVT GLPENVEEAI DHILNLEEEY LADVVDSEAL QVYMKPPAHE EAKAPSRGFV VRDAPWTASS SEKAPDMSSS EEFPSFGAQV APKTLPWGPK R (909-1268 aa encoded by BC001179)
High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDL-binding protein; High density lipoprotein-binding protein; lipoprotein-binding protein; predicted protein of HQ2900; unnamed protein product; Vigilin
Gene Aliases: 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDLBP; PRO2900; VGL
UniProt ID: (Human) Q00341, (Mouse) Q8VDJ3
Entrez Gene ID: (Human) 3069, (Mouse) 110611, (Rat) 64474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support