Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: AFNVKFSDVK IYKVGAPTIP DSCLPLGMSQ EDNQLKLVPV TPGRDMVHHL LSVSTAEGTE ENLSETSVAG FIVVTSVDLE HQVFTVLSPA PRPLPKNFLL IMDIRFMDLK
HEAB is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP/GTP-binding protein; CLP1; homolog of yeast CFIA subunit Clp1p; Polyadenylation factor Clp1; Polynucleotide kinase Clp1; Polyribonucleotide 5'-hydroxyl-kinase Clp1; Pre-mRNA cleavage complex II protein Clp1
Gene Aliases: CLP1; hClp1; HEAB
UniProt ID: (Human) Q92989
Entrez Gene ID: (Human) 10978
Molecular Function: mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support