Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • HEF1 Antibodies

          Invitrogen

          HEF1 Polyclonal Antibody

          View all (19) HEF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HEF1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          HEF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HEF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HEF1 Antibody (PA5-88292) in WB

          Western blot analysis of HEF1 in extracts of B-cell cells. Samples were incubated with HEF1 Polyclonal antibody (Product # PA5-88292) using a dilution of 1:3,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 10s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HEF1 Antibody in Western Blot (WB)
          HEF1 Polyclonal Antibody

          Product Details

          PA5-88292

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-154 of human NEDD9 (NP_8920112)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          4.3 mg/mL

          Amount

          430 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2804805

          Product Specific Information

          Immunogen sequence: MKYKNLMARA LYDNVPECAE ELAFRKGDIL TVIEQNTGGL EGWWLCSLHG RQGIVPGNRV KLLIGPMQET ASSHEQPASG LMQQTFGQQK LYQVPNPQAA PRDTIYQVPP SYQNQGIYQV PTGHGTQEQE VYQVPPSVQR SIGGTSGPHV GKKV; Positive Samples: B-cell; Cellular Location: Cell junction, Cell projection, Cytoplasm, Golgi apparatus, Nucleus, cell cortex, cytoskeleton, focal adhesion, lamellipodium, spindle

          Target Information

          HEF-1 is a multifunctional protein involved in integrin-based signaling that affects cell motility, growth, apoptosis and oncogenic transformation. The Cas family of docking proteins have been the subject of intense research because of their role in cell motility, growth, apoptosis and oncogenic transformation. These proteins are substrates of focal adhesion kinase (FAK) and the Src family of tyrosine kinases two active targets for drug development. HEF1 protein production increases levels of mRNA transcripts that encode proteins associated with motility, cell transformation and invasiveness, including several metalloproteinases, MLCK, p160ROCK and ErbBi. HEF1 overproduction also mediates apoptosis in epithelial-derived cell lines, including MCF7 and HeLa cells. Recent clinical studies at another institution have found that overexpression of BCAR1 (p130Cas), a related protein, is associated with tamoxifen resistance. This highlights the importance of studying the role of this family of proteins in cancer prognosis.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cas scaffolding protein family member 2; CAS-L; cas-like docking; CASS2; Crk-associated substrate related protein Cas-L; CRK-associated substrate-related protein; dJ49G10.2; dJ761I2.1; Enhancer of filamentation 1; Enhancer of filamentation 1 p55; hEF1; NEDD-9; Neural precursor cell expressed developmentally down-regulated protein 9; p105; p130Cas-related protein; pp105; p130Cas-related protein; Renal carcinoma antigen NY-REN-12; unnamed protein product

          View more View less

          Gene Aliases: CAS-L; CAS2; CASL; CASS2; HEF1; NEDD9

          View more View less

          UniProt ID: (Human) Q14511

          View more View less

          Entrez Gene ID: (Human) 4739

          View more View less

          Function(s)
          protein binding protein tyrosine kinase binding
          Process(es)
          cytoskeleton organization cell adhesion signal transduction transmembrane receptor protein tyrosine kinase signaling pathway integrin-mediated signaling pathway learning or memory cell migration positive regulation of cell migration negative regulation of cell migration regulation of actin cytoskeleton organization positive regulation of osteoclast differentiation actin filament bundle assembly cell division positive regulation of protein tyrosine kinase activity cilium disassembly lymphocyte migration into lymphoid organs positive regulation of lymphocyte chemotaxis positive regulation of substrate adhesion-dependent cell spreading positive regulation of dendritic spine maintenance positive regulation of cellular protein localization positive regulation of immunological synapse formation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline