Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HELLS Antibodies

          Abnova

          HELLS Polyclonal Antibody, MaxPab™

          View all (14) HELLS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HELLS Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          HELLS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HELLS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HELLS Antibody (H00003070-D01P) in WB

          Western Blot analysis of HELLS expression in transfected 293T cell line (H00003070-T01) by HELLS MaxPab polyclonal antibody.

          Lane 1: HELLS transfected lysate(97.10 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HELLS Antibody in Western Blot (WB)
          HELLS Polyclonal Antibody, MaxPab™

          Product Details

          H00003070-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HELLS (NP_060533.2, 1 a.a. approximately 838 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MPAERPAGSG GSEAPAMVEQ LDTAVITPAM LEEEEQLEAA GLERERKMLE KARMSWDRES TEIRYRRLQH LLEKSNIYSK FLLTKMEQQQ LEEQKKKEKL ERKKESLKVK KGKNSIDASE EKPVMRKKRG REDESYNISE VMSKEEILSV AKKNKKENED ENSSSTNLCV EDLQKNKDSN SIIKDRLSET VRQNTKFFFD PVRKCNGQPV PFQQPKHFTG GVMRWYQVEG MEWLRMLWEN GINGILADEM GLGKTVQCIA TIALMIQRGV PGPFLVCGPL STLPNWMAEF KRFTPDIPTM LYHGTQEERQ KLVRNIYKRK GTLQIHPVVI TSFEIAMRDR NALQHCYWKY LIVDEGHRIK NMKCRLIREL KRFNADNKLL LTGTPLQNNL SELWSLLNFL LPDVFDDLKS FESWFDITSL SETAEDIIAK EREQNVLHML HQILTPFLLR RLKSDVALEV PPKREVVVYA PLSKKQEIFY TAIVNRTIAN MFGSSEKETI ELSPTGRPKR RTRKSINYSK IDDFPNELEK LISQIQPEVD RERAVVEVNI PVESEVNLKL QNIMMLLRKC CNHPYLIEYP IDPVTQEFKI DEELVTNSGK FLILDRMLPE LKKRGHKVLL FSQMTSMLDI LMDYCHLRDF NFSRLDGSMS YSEREKNMHS FNTDPEVFIF LVSTRAGGLG INLTAADTVI IYDSDWNPQS DLQAQDRCHR IGQTKPVVVY RLVTANTIDQ KIVERAAAKR KLEKLIIHKN HFKGGQSGLN LSKNFLDPKE LMELLKSRDY EREIKGSREK VISDKDLELL LDRSDLIDQM NASGPIKEKM GIFKILENSE DSSPECLF

          Target Information

          This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: LSH; Lymphoid-specific helicase; Proliferation-associated SNF2-like protein; Senescence Associated Long Non-coding RNA; similar to Mus musculus lymphocyte specific helicase mRNA with GenBank Accession Number U25691.1; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; unnamed protein product

          View more View less

          Gene Aliases: HELLS; ICF4; LSH; Nbla10143; PASG; SALNR; SMARCA6

          View more View less

          UniProt ID: (Human) Q9NRZ9

          View more View less

          Entrez Gene ID: (Human) 3070

          View more View less

          Function(s)
          nucleotide binding chromatin binding helicase activity protein binding ATP binding hydrolase activity chromatin-protein adaptor activity ATP-dependent chromatin remodeler activity
          Process(es)
          double-strand break repair via homologous recombination urogenital system development kidney development double-strand break repair chromatin remodeling methylation-dependent chromatin silencing apoptotic process heterochromatin assembly pericentric heterochromatin assembly negative regulation of apoptotic process hypermethylation of CpG island lymphocyte proliferation cell division chromosomal DNA methylation maintenance following DNA replication cellular response to leukemia inhibitory factor negative regulation of intrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline