Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • HES1 Antibodies

          Invitrogen

          HES1 Polyclonal Antibody

          View all (46) HES1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HES1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          HES1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HES1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HES1 Antibody (PA5-88220) in WB

          Western blot analysis of HES1 in extracts of Rat testis. Samples were incubated with HES1 Polyclonal antibody (Product # PA5-88220) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 30s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HES1 Antibody in Western Blot (WB)
          HES1 Polyclonal Antibody

          Product Details

          PA5-88220

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 151-280 of human HES1 (NP_0055151)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          3.02 mg/mL

          Amount

          302 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2804753

          Product Specific Information

          Immunogen sequence: NAMTYPGQPH PALQAPPPPP PGPGGPQHAP FAPPPPLVPI PGGAAPPPGG APCKLGSQAG EAAKVFGGFQ VVPAPDGQFA FLIPNGAFAH SGPVIPVYTS NSGTSVGPNA VSPSSGPSLT ADSMWRPWRN; Positive Samples: Rat testis; Cellular Location: Nucleus

          Target Information

          This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ20408; Hairy and enhancer of split 1; Hairy-like; Hairy-like protein; RHL; Transcription factor HES-1

          View more View less

          Gene Aliases: Hes-1; Hes1; Hl

          View more View less

          UniProt ID: (Rat) Q04666

          View more View less

          Entrez Gene ID: (Rat) 29577

          View more View less

          Function(s)
          RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding transcription factor binding JUN kinase binding protein complex binding protein homodimerization activity histone deacetylase binding sequence-specific DNA binding chaperone binding E-box binding N-box binding basic helix-loop-helix transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development liver development embryonic heart tube morphogenesis outflow tract morphogenesis regulation of secondary heart field cardioblast proliferation ventricular septum development transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter protein complex assembly cell adhesion Notch signaling pathway smoothened signaling pathway STAT protein import into nucleus pattern specification process positive regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression negative regulation of neuron projection development response to organic cyclic compound cell migration telencephalon development midbrain-hindbrain boundary morphogenesis oculomotor nerve development trochlear nerve development hindbrain morphogenesis forebrain radial glial cell differentiation neural tube development pituitary gland development adenohypophysis development neuron differentiation lung development positive regulation of BMP signaling pathway midbrain development pancreas development somatic stem cell population maintenance hair cell differentiation aorta morphogenesis ascending aorta morphogenesis positive regulation of T cell proliferation auditory receptor cell differentiation positive regulation of tyrosine phosphorylation of Stat3 protein auditory receptor cell fate determination response to alkaloid positive regulation of DNA binding regulation of protein heterodimerization activity cell fate commitment negative regulation of cell differentiation regulation of fat cell differentiation negative regulation of auditory receptor cell differentiation negative regulation of neuron differentiation positive regulation of glial cell differentiation positive regulation of Notch signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of mitotic cell cycle, embryonic lateral inhibition positive regulation of JAK-STAT cascade cell maturation regulation of timing of cell differentiation thymus development cell morphogenesis involved in neuron differentiation positive regulation of astrocyte differentiation negative regulation of oligodendrocyte differentiation artery morphogenesis regulation of neurogenesis negative regulation of neurogenesis regulation of timing of neuron differentiation negative regulation of glial cell proliferation ventricular septum morphogenesis ureteric bud morphogenesis labyrinthine layer blood vessel development common bile duct development negative regulation of stomach neuroendocrine cell differentiation cardiac neural crest cell development involved in outflow tract morphogenesis pharyngeal arch artery morphogenesis cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to fatty acid glomerulus vasculature development comma-shaped body morphogenesis S-shaped body morphogenesis renal interstitial fibroblast development metanephric nephron tubule morphogenesis cochlea development negative regulation of calcium ion import response to thyroid hormone vascular smooth muscle cell development neuronal stem cell population maintenance negative regulation of RNA polymerase II regulatory region sequence-specific DNA binding cellular response to nerve growth factor stimulus negative regulation of pancreatic A cell differentiation negative regulation of stem cell differentiation negative regulation of pro-B cell differentiation negative regulation of forebrain neuron differentiation negative regulation of inner ear receptor cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline