Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • HIP1R Antibodies

          Abnova

          HIP1R Polyclonal Antibody, MaxPab™

          View all (13) HIP1R antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HIP1R Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          HIP1R Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HIP1R Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HIP1R Antibody (H00009026-D01P) in WB

          Western Blot analysis of HIP1R expression in transfected 293T cell line (H00009026-T02) by HIP1R MaxPab polyclonal antibody.

          Lane 1: HIP1R transfected lysate(119.40 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HIP1R Antibody in Western Blot (WB)
          HIP1R Antibody in Western Blot (WB)
          HIP1R Antibody in Western Blot (WB)
          HIP1R Polyclonal Antibody, MaxPab™

          Product Details

          H00009026-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HIP1R (NP_003950.1, 1 a.a. approximately 1068 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MNSIKNVPAR VLSRRPGHSL EAEREQFDKT QAISISKAIN TQEAPVKEKH ARRIILGTHH EKGAFTFWSY AIGLPLPSSS ILSWKFCHVL HKVLRDGHPN VLHDCQRYRS NIREIGDLWG HLHDRYGQLV NVYTKLLLTK ISFHLKHPQF PAGLEVTDEV LEKAAGTDVN NIFQLTVEMF DYMDCELKLS ESVFRQLNTA IAVSQMSSGQ CRLAPLIQVI QDCSHLYHYT VKLLFKLHSC LPADTLQGHR DRFHEQFHSL RNFFRRASDM LYFKRLIQIP RLPEGPPNFL RASALAEHIK PVVVIPEEAP EDEEPENLIE ISTGPPAGEP VVVADLFDQT FGPPNGSVKD DRDLQIESLK REVEMLRSEL EKIKLEAQRY IAQLKSQVNA LEGELEEQRK QKQKALVDNE QLRHELAQLR AAQLEGERSQ GLREEAERKA SATEARYNKL KEKHSELVHV HAELLRKNAD TAKQLTVTQQ SQEEVARVKE QLAFQVEQVK RESELKLEEK SDQLEKLKRE LEAKAGELAR AQEALSHTEQ SKSELSSRLD TLSAEKDALS GAVRQREADL LAAQSLVRET EAALSREQQR SSQEQGELQG RLAERESQEQ GLRQRLLDEQ FAVLRGAAAE AAGILQDAVS KLDDPLHLRC TSSPDYLVSR AQEALDAVST LEEGHAQYLT SLADASALVA ALTRFSHLAA DTIINGGATS HLAPTDPADR LIDTCRECGA RALELMGQLQ DQQALRHMQA SLVRTPLQGI LQLGQELKPK SLDVRQEELG AVVDKEMAAT SAAIEDAVRR IEDMMNQARH ASSGVKLEVN ERILNSCTDL MKAIRLLVTT STSLQKEIVE SGRGAATQQE FYAKNSRWTE GLISASKAVG WGATQLVEAA DKVVLHTGKY EELIVCSHEI AASTAQLVAA SKVKANKHSP HLSRLQECSR TVNERAANVV ASTKSGQEQI EDRDTMDFSG LSLIKLKKQE METQVRVLEL EKTLEAERMR LGELRKQHYV LAGASGSPGE EVAIRPSTAP RSVTTKKPPL AQKPSVAPRQ DHQLDKKDGI YPAQLVNY

          Target Information

          Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HIP-12; HIP1-related protein; huntingtin interacting protein 12; Huntingtin-interacting protein 1-related protein; Huntingtin-interacting protein 12; unnamed protein product

          View more View less

          Gene Aliases: AA410023; HIP12; HIP1R; HIP3; ILWEQ; KIAA0655; mKIAA0655

          View more View less

          UniProt ID: (Human) O75146, (Mouse) Q9JKY5

          View more View less

          Entrez Gene ID: (Human) 9026, (Mouse) 29816

          View more View less

          Function(s)
          actin binding protein binding phospholipid binding phosphatidylinositol-4,5-bisphosphate binding phosphatidylinositol-3,4,5-trisphosphate binding SH3 domain binding clathrin binding clathrin light chain binding phosphatidylinositol binding clathrin adaptor activity identical protein binding protein homodimerization activity phosphatidylinositol-3,4-bisphosphate binding protein heterodimerization activity actin filament binding phosphatidylinositol-3,5-bisphosphate binding non-motor actin binding protein
          Process(es)
          endocytosis receptor-mediated endocytosis actin filament organization regulation of endocytosis negative regulation of actin filament polymerization regulation of actin cytoskeleton organization negative regulation of Arp2/3 complex-mediated actin nucleation regulation of apoptotic process negative regulation of apoptotic process positive regulation of epidermal growth factor receptor signaling pathway clathrin coat assembly protein stabilization positive regulation of transport positive regulation of cellular component organization digestive system development regulation of gastric acid secretion membrane organization apoptotic signaling pathway intrinsic apoptotic signaling pathway postsynapse organization positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway positive regulation of clathrin coat assembly regulation of clathrin-mediated endocytosis positive regulation of platelet-derived growth factor receptor-beta signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline