Recommended positive controls: HepG2, human ovary tumor.
Predicted reactivity: Zebrafish: 92%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HLX is a transcription factor required for TBX21/T-bet-dependent maturation of Th1 cells as well as maintenance of Th1-specific gene expression. Involved in embryogenesis and hematopoiesis.