Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAAMRKALP RRLVGLASLR AVSTSSMGTL PKRVKIVEVG PRDGLQNEKN IVSTPVKIKL IDMLSEAGLS VIETTSFVSP KWVPQMGDHT EVLKGIQKFP GINYPVLTPN LKGFEAAVAA GAKEVVIFGA ASELFTKKNI NCSIEESFQR FDAILKAAQS ANISVRGYVS CALGCPYEGK ISPAKVAEVT KKFYSMGCYE ISLGDTIGVG TPGIMKDMLS AVMQEVPLAA LAVHCHDTYG QALANTLMAL QMGVSVVDSS VAGLGGCPYA QGASGNLATE DLVYMLEGLG IHTGVNLQKL LEAGNFICQA LNRKTSSKVA QATCKL (1-325 aa encoded by B C010570)
The protein encoded by this gene belongs to the HMG-CoA lyase family. It is a mitochondrial enzyme that catalyzes the final step of leucine degradation and plays a key role in ketone body formation. Mutations in this gene are associated with HMG-CoA lyase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxy-3-methylglutarate-CoA lyase; 3-hydroxy-3-methylglutaryl-Coenzyme A lyase; 3-hydroxymethyl-3-methylglutaryl-CoA lyase; 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase; HL; HMG-CoA lyase; HMGCL; hydroxymethylglutaricaciduria; mitochondrial 3-hydroxy-3-methylglutaryl-CoA lyase; unnamed protein product
Gene Aliases: HL; HMGCL1
UniProt ID: (Human) P35914, (Rat) P97519, (Mouse) P38060
Entrez Gene ID: (Human) 3155, (Rat) 79238, (Mouse) 15356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support