Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TCF2 Antibodies

          Proteintech

          HNF1B Polyclonal Antibody

          26 References
          View all (30) TCF2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HNF1B Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          HNF1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HNF1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HNF1B Antibody (12533-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 4% PFA ) fixed L02 cells using 12533-1-AP (HNF1B antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HNF1B Antibody in Immunocytochemistry (ICC/IF)
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNF1B Antibody in Western Blot (WB)
          HNF1B Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          HNF1B Polyclonal Antibody

          Product Details

          12533-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 11 publications 11 publications

          Immunohistochemistry (IHC)

          -
          View 10 publications 10 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 3 publications 3 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Pig

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HNF1B Fusion Protein Ag3232 (1-310 aa encoded by BC017714)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.28 mg/mL

          Amount

          42 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This is a rabbit polyclonal antibody raised against the N-terminal region of human HNF1B. HNF1B exists some isoforms with MW of 61 kDa, 58 kDa, 46 kDa, and 44 kDa.

          Immunogen sequence: MVSKLTSLQ QELLSALLSS GVTKEVLVQA LEELLPSPNF GVKLETLPLS PGSGAEPDTK PVFHTLTNGH AKGRLSGDEG SEDGDDYDTP PILKELQALN TEEAAEQRAE VDRMLSEDPW RAAKMIKGYM QQHNIPQREV VDVTGLNQSH LSQHLNKGTP MKTQKRAALY TWYVRKQREI LRQFNQTVQS SGNMTDKSSQ DQLLFLFPEF SQQSHGPGQS DDACSEPTNK KMRRNRFKWG PASQQILYQA YDRQKNPSKE EREALVEECN RAECLQRGVS PSKAHGLGSN LVTEVRVYNW FANRRKEEAF R (1-310 aa encoded by BC017714)

          Target Information

          Hepatocyte Nuclear Factor 1 (HNF1, Homeoprotein LFB3, Transcription factor 2, TCF2, Variant hepatic nuclear factor) is a member of the homeodomain-containing superfamily of transcription factors. It is a liver-specific factor of the homeobox-containing basic helix-turn-helix family. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. HNF1has been shown to function in nephron development and regulates development of the embryonic pancreas. Mutations in the gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of the gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HNF1 beta A; homeoprotein LFB3; TCF2; Tcf2-splice variant; transcription factor; transcription factor 2; Transcription factor 2, hepatic; unnamed protein product; variant hepatic nuclear factor 1

          View more View less

          Gene Aliases: ADTKD3; FJHN; HNF-1-beta; HNF-1B; HNF-1Beta; HNF1beta; HNF2; HPC11; LF-B3; LFB3; MODY5; RCAD; T2D; TCF-2; TCF2; VHNF1

          View more View less

          UniProt ID: (Human) P35680, (Mouse) P27889, (Rat) P23899

          View more View less

          Entrez Gene ID: (Human) 6928, (Mouse) 21410, (Rat) 25640

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcription cofactor binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding identical protein binding protein homodimerization activity promoter-specific chromatin binding sequence-specific DNA binding macromolecular complex binding double-stranded DNA binding
          Process(es)
          kidney development liver development regulation of transcription from RNA polymerase II promoter insulin secretion pancreas development endocrine pancreas development regulation of pronephros size pronephric nephron tubule development positive regulation of transcription, DNA-templated animal organ development system development pronephros development genitalia development positive regulation of transcription initiation from RNA polymerase II promoter negative regulation of transcription from RNA polymerase II promoter formation of primary germ layer endoderm formation endodermal cell fate specification blastocyst development inner cell mass cell differentiation Notch signaling pathway endoderm development response to glucose anterior/posterior pattern specification gene expression positive regulation of gene expression regulation of Wnt signaling pathway hindbrain development circadian regulation of gene expression negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter embryonic digestive tract morphogenesis embryonic morphogenesis branching morphogenesis of an epithelial tube epithelial cell proliferation epithelium development ureteric bud elongation kidney morphogenesis hepatoblast differentiation protein-DNA complex assembly regulation of branch elongation involved in ureteric bud branching mesonephric tubule development nephric duct development mesonephric duct development nephric duct formation mesonephric duct formation negative regulation of mesenchymal cell apoptotic process involved in metanephros development response to xenobiotic stimulus response to carbohydrate hepatocyte differentiation response to biphenyl
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-kzhm8:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline