Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSEEQFGGD GAAAAATAAV GGSAGEQEGA MVAATQGAAA AAGSGAGTGG GTASGGTEGG SAESEGAKID ASKNEEDEGH SNSSPRHSEA ATAQREEWKM FIGGLSWDTT KKDLKDYFSK FGEVVDCTLK LDPITGRSRG FGFVLFKESE SVDKVMDQKE HKLNGKVIDP KRAKAMKTKE PVKKIFVGGL SPDTPEEKIR EYFGGFGEVE SIELPMDNKT NKRRGFCFIT FKEEEPVKKI MEKKYHNVGL SKCEIKVAMS KEQYQQQQQW GSRGGFAGRA RGRGGDQQSG YGKVSRRGGH QNSYKPY (1-306 aa encoded by BC026015)
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are nucleic acid binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. It localizes to both the nucleus and the cytoplasm. This protein is implicated in the regulation of mRNA stability. Alternative splicing of this gene results in four transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARE-binding protein AUFI, type A; AU-rich element RNA binding protein 1, 37kDa; AU-rich element RNA-binding factor 1; AU-rich element RNA-binding protein 1; AU-rich RNA-binding factor 1; hnRNP D0; RNA binding protein p45AUF1; transcription factor; unnamed protein product
Gene Aliases: AUF1; AUF1A; hnRNPD0; HNRPD; P37
UniProt ID: (Human) Q14103, (Mouse) Q60668, (Rat) Q9JJ54
Entrez Gene ID: (Human) 3184, (Mouse) 11991, (Rat) 79256
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support