Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • hnRNP U Antibodies

          Proteintech

          HNRNPU Polyclonal Antibody

          18 References
          View all (14) hnRNP U antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HNRNPU Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          HNRNPU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HNRNPU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HNRNPU Antibody (14599-1-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 14599-1-AP (HNRNPU antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HNRNPU Antibody in Immunocytochemistry (ICC/IF)
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Immunoprecipitation (IP)
          HNRNPU Polyclonal Antibody

          Product Details

          14599-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 14 publications 14 publications

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HNRNPU Fusion Protein Ag6141 (10-236 aa encoded by BC003621)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.62 mg/mL

          Amount

          93 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: KLKVSELKE ELKKRRLSDK GLKAELMERL QAALDDEEAG GRPAMEPGNG SLDLGGDSAG RSGAGLEQEA AAGGDEEEEE EEEEEEGISA LDGDQMELGE ENGAAGAADS GPMEEEEAAS EDENGDDQGF QEGEDELGDE EEGAGDENGH GEQQPQPPAT QQQQPQQQRG AAKEAAGKSS GPTSLFAVTV APPGARQGQQ QAGGDGKTEQ KGGDKKRGVK RPREDHGR (10-236 aa encoded by BC003621 )

          Target Information

          The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: contains signal peptide domain; FLJ30202; FLJ37978; GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; hnRNP U; HNRNPU antisense RNA 1; NCRNA00201; nuclear matrix protein sp120; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; SAF-A; SAR-DNA binding protein of the nuclear scaffold; scaffold attachment factor A; Scaffold-attachment factor A; SP120; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U211; unnamed protein product

          View more View less

          Gene Aliases: AA408410; AI256620; AL024194; AL024437; AW557595; C1orf199; C86794; DEE54; EIEE54; GRIP120; hnRNP U; HNRNPU; HNRNPU-AS1; HNRPU; pp120; rCG_20317; SAF-A; SAFA; SN1; Sp120; U21.1

          View more View less

          UniProt ID: (Human) Q00839, (Rat) Q6IMY8, (Mouse) Q8VEK3

          View more View less

          Entrez Gene ID: (Human) 3192, (Rat) 117280, (Mouse) 51810

          View more View less

          Function(s)
          nucleotide binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core binding TFIIH-class transcription factor binding DNA binding chromatin binding double-stranded DNA binding single-stranded DNA binding transcription corepressor activity RNA binding double-stranded RNA binding single-stranded RNA binding mRNA 3'-UTR binding actin binding protein binding ATP binding poly(A) binding snRNA binding poly(C) RNA binding chromatin DNA binding poly(G) binding piRNA binding pre-mRNA binding identical protein binding ribonucleoprotein complex binding sequence-specific DNA binding macromolecular complex binding telomerase RNA binding RNA polymerase II C-terminal domain binding lncRNA binding sequence-specific double-stranded DNA binding promoter-specific chromatin binding core promoter binding transporter activity poly(A) RNA binding
          Process(es)
          osteoblast differentiation transport circadian regulation of gene expression CRD-mediated mRNA stabilization negative regulation of transcription from RNA polymerase II promoter alternative mRNA splicing, via spliceosome regulation of alternative mRNA splicing, via spliceosome mRNA splicing, via spliceosome chromatin organization RNA processing mRNA processing regulation of mitotic cell cycle RNA splicing dosage compensation by inactivation of X chromosome regulation of gene expression positive regulation of macromolecule biosynthetic process mRNA metabolic process cell differentiation erythrocyte differentiation chromatin silencing by small RNA negative regulation of telomere maintenance via telomerase negative regulation of kinase activity negative regulation of transcription elongation from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter mRNA stabilization rhythmic process cell division maintenance of protein location in nucleus cardiac muscle cell development random inactivation of X chromosome cellular response to glucocorticoid stimulus positive regulation of brown fat cell differentiation dendritic transport of messenger ribonucleoprotein complex negative regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay regulation of mitotic spindle assembly regulation of chromatin organization positive regulation of attachment of mitotic spindle microtubules to kinetochore protein localization to spindle microtubule RNA localization to chromatin cellular response to leukemia inhibitory factor adaptive thermogenesis positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity positive regulation of stem cell proliferation negative regulation of stem cell differentiation positive regulation of cytoplasmic translation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline