Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MVGGGGKRRP GGEGPQCEKT TDVKKSKFCE ADVSSDLRKE VENHYKLSLP EDFYHFWKFC EELDPEKPSD SLSASLGLQL VGPYDILAGK HKTKKKSTGL NFNLHWRFYY DPPEFQTIII GDNKTQYH
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C4orf27; Chromosome 4 Open Reading Frame 27; FLJ20534; Histone PARylation factor 1; HPF1; unnamed protein product; UPF0609 protein C4orf27
Gene Aliases: C4orf27; HPF1
UniProt ID: (Human) Q9NWY4
Entrez Gene ID: (Human) 54969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support