Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: AVLVDYIRV LDFDVKRKYF RQELERLDEG LRKEDMAVHV LRDHVFEDSY RELHRKSPEE MKNRLYIVFE GEEGQDAGGL LREWYMIISR EMFNPMYALF RTSPGDRVTY TINPSSHCNP NHLSYFKFVG RIVAKAVYDN RLLECYFTRS FYKHILGKSV RYTDMESEDY HFYQGLVYLL ENDVSTLGYD LTFSTEVQEF GVCEVRDLKP NGANILVTEE NKKEYVHLVC QMRMTGAIRK QLAAFLEGFY EIIPKRLISI FTEQELELLI SGLPTIDIDD LKSNTEYHKY QSNSIQIQWF WRALRSFDQA DRAKFLQFVT GTSKVPLQGF AALEGMNGIQ KFQIHRDDRS TDRLPSAHTC FNQLDLPAYE SFEKLRHMLL LAIQECSEGF GLA (3983-4374 aa encoded by BC002602)
SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARF-binding protein 1; BJ-HCC-24 tumor antigen; DNA binding protein; E3Histone; HECT domain protein LASU1; HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase; HECT, UBA and WWE domain-containing protein 1; HECT-type E3 ubiquitin transferase HUWE1; homologous to E6AP carboxyl terminus homologous protein 9; KIAA0312; large structure of URE B1; large structure of UREB1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; MULE; RP3-339A18.4; ubiquitin ligase with HECT domain; unnamed protein product; upstream regulatory element binding protein 1; upstream regulatory element-binding protein 1; URE-B1; URE-binding protein 1; UREB1
Gene Aliases: 5430439H10Rik; ARF-BP1; C430014N20Rik; Gm1718; HECTH9; HSPC272; Ib772; LASU1; MRXST; MULE; URE-B1; UREB1
UniProt ID: (Human) Q7Z6Z7, (Mouse) Q7TMY8
Entrez Gene ID: (Human) 10075, (Mouse) 59026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support