    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Influenza B NP Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          Influenza B NP Monoclonal Antibody (1D8)
          View all (18) Influenza B NP antibodies
          Tech Support
          Cite Influenza B NP Monoclonal Antibody (1D8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Influenza B NP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Influenza B NP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Influenza B NP Antibody (HYB 116-01-02) in ICC/IF
          Immunofluorescent analysis of Influenza B NP in infected Vero cells using an Influenza B NP monoclonal antibody (Product # HYB 116-01-02) followed by detection using a secondary polyclonal rabbit anti-mouse Ig FITC conjugated antibody at a dilution of 1:1000. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Influenza B NP Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          HYB 116-01-02
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:1,000
          -
          ELISA (ELISA)
          1:200
          -
          Product Specifications
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D8
          Immunogen
          Influenza B virus.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.04 mg/mL
          Amount
          208 µg
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          RRID
          AB_1076169
          Product Specific Information
          HYB 116-01-02 detects the nucleoprotein (NP) of Influenza B virus. This antibody does not cross react with the the Influenza A virus in patient samples or purified Influenza A virus.
          HYB 116-01-02 has been successfully used in Western blotting, ELISA and immunofluorescence procedures. By ELISA, HYB 116-01-02 is suitable as capture antibody in combination with HYB 116-03-02 as detecting antibody.
          HYB 116-01-02 reacts with Influenza B virus, acetone-fixed Vero cells when diluted 1:1,000. Specific binding can be visualized by immunofluorescence using a polyclonal anti-mouse Ig antibody labeled with, for example, florescein.
          The HYB 116-01-02 immunogen is Influenza B virus.
          Epitope of HYB 116-01-02 differs from that of HYB 156-02-02.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
          Target Information
          Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline