    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • MBL2 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          MBL2 Monoclonal Antibody (3B6), Biotin
          3 Published Figures
          3 References
          View all (19) MBL2 antibodies
          Tech Support
          Cite MBL2 Monoclonal Antibody (3B6), Biotin
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (3)
          MBL2 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          MBL2 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          MBL2 Antibody (HYB 131-01B-005) in ELISA
          Sandwich ELISA of Mannan-Binding Lectin using HYB 131-01 as the capture antibody and HYB 131-01B as the biotinylated detection antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MBL2 Antibody in ELISA (ELISA)
          MBL2 Antibody in Immunocytochemistry (ICC/IF)
          MBL2 Antibody in Flow Cytometry (Flow)
          MBL2 Antibody in Dot Blot (DB)
          Product Details
          HYB 131-01B-005
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          -
          View 1 publication 1 publication
          Flow Cytometry (Flow)
          -
          View 1 publication 1 publication
          ELISA (ELISA)
          1:40,000
          -
          Dot blot (DB)
          -
          View 1 publication 1 publication
          Product Specifications
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3B6
          Immunogen
          MBL purified from human donor plasma
          Conjugate
          Biotin Biotin Biotin
          Form
          Liquid
          Concentration
          0.92 mg/mL
          Amount
          46 µg
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          RRID
          AB_1076464
          Product Specific Information
          HYB 131-01B-005 is specific for MBL from human serum or plasma.
          HYB 131-01B-005 has been successfully used in ELISA. Used as detection antibody in sandwich ELISA with a mannan or HYB 131-01B-005 coat, HYB 131-01B-005 is selective for normally oligomerized MBL.
          The HYB 131-01B-005 immunogen is MBL purified from human donor plasma.
          Epitope specificity differs from that of HYB 131-10-02 and HYB 131-11-02. Epitope is near the carbohydrate-binding site.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
          Target Information
          MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline