    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • MBL2 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          MBL2 Monoclonal Antibody (15C5)
          1 Published Figure
          1 Reference
          View all (21) MBL2 antibodies
          Tech Support
          Cite MBL2 Monoclonal Antibody (15C5)
          Additional Information:
          {{banner.description}}
          • Published Figures (1)
          MBL2 Antibody in Dot Blot (DB)
          Group 53 Created with Sketch.
          MBL2 Antibody in Dot Blot (DB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MBL2 Antibody (HYB 131-14-02) in DB
          PloS one 2017 - Published figure using MBL2 monoclonal antibody (Product # HYB 131-14-02) in Dot Blot {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MBL2 Antibody in Dot Blot (DB)
          Product Details
          HYB 131-14-02
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          ELISA (ELISA)
          1:250
          -
          Dot blot (DB)
          -
          View 1 publication 1 publication
          Product Specifications
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          15C5
          Immunogen
          Human MBL.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.91 mg/mL
          Amount
          182 µg
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Wet ice
          RRID
          AB_1076470
          Product Specific Information
          HYB 131-14-02 is specific for human MBL from plasma or serum, but cross-reacts strongly with MBL from horse and swine.
          HYB 131-14-02 has been successfully used in ELISA and Western blotting procedures. HYB 131-14-02 reacts strongly with horse and swine serum, when captured on an mannan coat in ELISA.
          The HYB 131-14-02 immunogen is human MBL.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
          Target Information
          MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline