PA1-25689 detects Haptoglobin from human, mouse, and rat samples.
PA1-25689 has been successfully used in Western blot and ELISA procedures. The predicted molecular weight is 45 kDa.
The PA1-25689 immunogen is human plasma protein.
Store at 4°C short term. For extended storage aliquot and store at -20°C or below, avoiding freeze/thaw cycles.
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which also acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chains bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.