Search Thermo Fisher Scientific
- Contact Us
- Quick Order
Invitrogen
MA1-7612 detects HCV from virus samples.
MA1-7612 has been successfully used in ELISA, Western blot and immunoprecipitation procedures.
The MA1-7612 immunogen is a purified recombinant HCV core 00115V protein.
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.