Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Histone H2A Antibodies

          Active Motif

          Histone H2A, acidic patch Polyclonal Antibody

          4 References
          View all (30) Histone H2A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Histone H2A, acidic patch Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          Histone H2A, acidic patch Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Histone H2A, acidic patch Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Histone H2A, acidic patch Antibody (39111) in WB

          Histone H2A, acidic patch pAb tested by Western blot. Detection of Histone H2A by Western blot. The analysis was performed using recombinant histone H2A protein (200 ng, lane 1) and acid extract of HeLa cells (5 µg, lane 2) probed with Histone H2A, acidic patch pAb (1:1,000 dilution). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Histone H2A, acidic patch Antibody in Western Blot (WB)
          Histone H2A, acidic patch Antibody in ChIP Assay (ChIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Histone H2A, acidic patch Polyclonal Antibody

          Product Details

          39111

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          ChIP assay (ChIP)

          10 µL
          -

          ChIP-sequencing (ChIP-Seq)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          This Histone H2A, acidic patch antibody was raised against a peptide including amino acids 89-91 of histone H2A.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum with 30% glycerol

          Contains

          0.035% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2793159

          Target Information

          Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Acetyl-Histone H2; buforin I; H2A histone family member; H2A histone family member J; H2A histone family member V; H2A histone family member X; H2A histone family member Z; H2A histone family, member A; H2A histone family, member C; H2A histone family, member D; H2A histone family, member E; H2A histone family, member G; H2A histone family, member I; H2A histone family, member L; H2A histone family, member M; H2A histone family, member N; H2A histone family, member O; H2A histone family, member P; H2A histone family, member Q; H2A histone family, member R; H2A.1; H2A.F/Z; H2A.W histone; H2A/d; H2a/j; h2ad; H2ae; H2AFX; H2AFZ; h2ai; H2ak; H2AX histone; H2AZ histone; HIST1H3D; HIST2H2AA; HIST3H2A; histone (H2A.Z); histone 1, H2aa; histone 1, H2ab; histone 1, H2ac; histone 1, H2ad; histone 1, H2ae; histone 1, H2ag; histone 1, H2ai; histone 1, H2aj; histone 1, H2ak; histone 1, H2al; histone 1, H2am; histone 2, H2aa3; histone 2, H2aa4; histone 2, H2ab; histone 2, H2ac; Histone 2A; histone 3, H2a; histone cluster 1 H2A family member a; histone cluster 1 H2A family member b; histone cluster 1 H2A family member c; histone cluster 1 H2A family member d; histone cluster 1 H2A family member e; histone cluster 1 H2A family member g; histone cluster 1 H2A family member h; histone cluster 1 H2A family member i; histone cluster 1 H2A family member j; histone cluster 1 H2A family member k; histone cluster 1 H2A family member l; histone cluster 1 H2A family member m; histone cluster 1, H2aa; histone cluster 1, H2ab; histone cluster 1, H2ac; histone cluster 1, H2ad; histone cluster 1, H2ae; histone cluster 1, H2ag; histone cluster 1, H2ah; histone cluster 1, H2ai; histone cluster 1, H2ak; histone cluster 1, H2al; histone cluster 1, H2am; histone cluster 2 H2A family member a3; histone cluster 2 H2A family member a4; histone cluster 2 H2A family member b; histone cluster 2 H2A family member c; histone cluster 2, H2aa3; histone cluster 2, H2aa4; histone cluster 2, H2ab; histone cluster 2, H2ac; histone cluster 3 H2A; histone cluster 3, H2a; histone H2A type 1-J; histone H2A-GL101; histone H2A.3; histone H2A.F/Z; histone H2A.x; histone H2A/a; histone H2A/g; histone H2A/l; histone H2A/m; histone H2A/p; histone H2A/ptl; histone H2A/q; histone H2A/r; histone H2A/s; histone H2AC; histone H2AD; histone H2AE; histone IIa; purine-rich binding element protein B; unnamed protein product

          View more View less

          Gene Aliases: bA317E16.2; dJ160A22.4; dJ193B12.1; dJ193B12.9; dJ221C16.4; dJ86C11.1; H2A; H2a-615; H2A-GL101; H2A.1; H2A.1b; H2A.2; H2A.3; H2A.i; H2A.X; H2A.z; H2A.Z-1; H2A.Z-2; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/i; H2A/l; H2A/m; H2A/n; H2A/O; H2A/p; H2A/q; H2A/R; H2A/S; H2A/X; H2A/z; H2AA; H2AB; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFJ; H2AFL; H2AFM; H2AFN; H2AFO; H2AFP; H2AFQ; H2AFR; H2AFV; H2AFX; H2AFZ; H2AG; H2AH; H2AV; H2AW; H2AZ; HIST1H2AA; HIST1H2AB; HIST1H2AC; HIST1H2AD; HIST1H2AE; HIST1H2AG; HIST1H2AH; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; HIST2H2AA; HIST2H2AA3; HIST2H2AA4; HIST2H2AB; HIST2H2AC; HIST3H2A; HISTH2AA; pH2A/f; TH2A

          View more View less

          UniProt ID: (Human) Q96QV6, (Human) P0C0S8, (Human) Q96KK5, (Human) Q99878, (Human) Q6FI13, (Human) Q16777, (Human) Q8IUE6, (Human) Q7L7L0, (Human) P04908, (Human) Q93077, (Human) P20671, (Human) Q9BTM1, (Human) P16104, (Human) P0C0S5, (Human) Q71UI9

          View more View less

          Entrez Gene ID: (Human) 221613, (Human) 8969, (Human) 85235, (Human) 8329, (Human) 8331, (Human) 8330, (Human) 8332, (Human) 8336, (Human) 8337, (Human) 723790, (Human) 8338, (Human) 317772, (Human) 92815, (Human) 8335, (Human) 8334, (Human) 3013, (Human) 3012, (Human) 55766, (Human) 3014, (Human) 3015, (Human) 94239

          View more View less

          Function(s)
          DNA binding structural constituent of chromatin protein heterodimerization activity protein binding enzyme binding damaged DNA binding histone binding chromatin-protein adaptor activity RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding chromatin DNA binding nucleosomal DNA binding binding, bridging
          Process(es)
          chromatin organization negative regulation of cell proliferation heterochromatin assembly protein localization to CENP-A containing chromatin DNA damage checkpoint double-strand break repair nucleosome assembly cellular response to DNA damage stimulus response to ionizing radiation positive regulation of DNA repair protein localization to site of double-strand break nucleosome disassembly UV-damage excision repair positive regulation of transcription from RNA polymerase II promoter cellular response to estradiol stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0