Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: QYINLQLLNA KPQECLMGTV GTLLLENPLG QNGLTHQGLL YEAAKVFGLR SRKLKLFLNE TQTQEITEDI PVKTLNMKTV YVSVLPTTAD F
IARS is a member of aminoacyl-tRNA synthetases which catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Isoleucine-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family and has been identified as a target of autoantibodies in the autoimmune disease polymyositis/dermatomyositis. Two alternatively spliced variants have been isolated that represent alternate 5' UTRs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20736; IRS; isoleucine tRNA ligase 1, cytoplasmic; Isoleucine--tRNA ligase, cytoplasmic; Isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase, cytoplasmic
Gene Aliases: IARS; IARS1; ILERS; ILRS; IRS; PRO0785
UniProt ID: (Human) P41252
Entrez Gene ID: (Human) 3376
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support