Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TQKEDDCDE IQCQLAIPVS SLNSQYCVSA EGVLHVWGVT TEKSKEVCIT IFNSSIKGSL WIPVVAALLL FLVLSLVFIC FYIKKINPLK EKSIILPKSL ISVVRSATLE TKPESKYVSL ITSYQPFSLE KEVVCEEPLS PATVPGMHTE DNPGKVEHTE ELSSITEVVT TEENIPDVVP GSHLTPIERE SSSPLSSNQS EPGSIALNSY HSRNCSESDH SRNGFDTDSS CLESHSSLSD SEFPPNNKGE IKTEGQELIT VIKAPTSFGY DKPHVLVDLL VDDSGKESLI GYRPTEDSKE FS (189-489 aa encoded by BC005333)
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antiviral protein, type 2; AVP, type 2; CD119 antigen; CDw119; cytokine receptor; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; IFNgR1; immune interferon receptor 1; interferon-gamma receptor; interferon-gamma receptor alpha chain; unnamed protein product
Gene Aliases: CD119; IFNGR; IMD27A; IMD27B
UniProt ID: (Human) P15260
Entrez Gene ID: (Human) 3459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support