Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • IFT140 Antibodies

          Proteintech

          IFT140 Polyclonal Antibody

          77 References
          View all (4) IFT140 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IFT140 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          IFT140 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          IFT140 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IFT140 Antibody (17460-1-AP) in ICC/IF

          IF result (enrichment to the base (arrowhead) and tip (asterix) of cilia) of anti-IFT140 (17460-1-AP, 1:50) with serum-starved hTERT-RPE1 (PFA fixed) by Dr. Moshe Kim. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IFT140 Antibody in Immunocytochemistry (ICC/IF)
          IFT140 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT140 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT140 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT140 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT140 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          IFT140 Antibody in Western Blot (WB)
          IFT140 Antibody in Western Blot (WB)
          IFT140 Antibody in Western Blot (WB)
          IFT140 Antibody in Immunoprecipitation (IP)
          IFT140 Polyclonal Antibody

          Product Details

          17460-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 19 publications 19 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 51 publications 51 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IFT140 Fusion Protein Ag11119 (1114-1462 aa encoded by BC035577)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.49 mg/mL

          Amount

          73.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: AEDLDETSD PALLARCSDF FIEHSQYERA VELLLAARKY QEALQLCLGQ NMSITEEMAE KMTVAKDSSD LPEESRRELL EQIADCCMRQ GSYHLATKKY TQAGNKLKAM RALLKSGDTE KITFFASVSR QKEIYIMAAN YLQSLDWRKE PEIMKNIIGF YTKGRALDLL AGFYDACAQV EIDEYQNYDK AHGALTEAYK CLAKAKAKSP LDQETRLAQL QSRMALVKRF IQARRTYTED PKESIKQCEL LLEEPDLDST IRIGDVYGFL VEHYVRKEEY QTAYRFLEEM RRRLPLANMS YYVSPQAVDA VHRGLGLPLP RTVPEQVRHN SMEDARELDE EVVEEADDDP (1114-1462 aa encoded by BC035577)

          Target Information

          IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: intraflagellar transport 140; intraflagellar transport 140 homolog; Intraflagellar transport protein 140 homolog; T-complex expressed gene 5; unnamed protein product; WD and tetratricopeptide repeats 2; WD and tetratricopeptide repeats protein 2

          View more View less

          Gene Aliases: AI661311; c305C8.4; c380F5.1; CED5; gs114; IFT140; KIAA0590; mKIAA0590; MZSDS; PKD9; RP80; SRTD9; Tce5; WDTC2

          View more View less

          UniProt ID: (Human) Q96RY7, (Mouse) E9PY46

          View more View less

          Entrez Gene ID: (Human) 9742, (Mouse) 106633, (Rat) 100362124

          View more View less

          Function(s)
          protein binding molecular_function structural protein
          Process(es)
          determination of left/right symmetry heart development regulation of smoothened signaling pathway neural tube patterning cell projection organization embryonic camera-type eye development limb morphogenesis intraciliary retrograde transport photoreceptor cell outer segment organization intraciliary transport embryonic digit morphogenesis embryonic cranial skeleton morphogenesis cilium morphogenesis protein localization to cilium regulation of cilium assembly non-motile cilium assembly embryonic brain development cilium assembly skeletal system morphogenesis retina development in camera-type eye renal system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0